Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C23H37N5O10
Molecular Weight:
543.57
NACRES:
NA.32
UNSPSC Code:
12352200
assay
≥95% (HPLC)
form
lyophilized
composition
Peptide Content, ≥80%
storage condition
protect from light
storage temp.
−20°C
Application
Caspase 2 Inhibitor (Ac-VDVAD-CHO) is a non-selective (also inhibits caspase 3) but powerful inhibitor of caspase 2 used to study the role of caspase 2 in cell processes.
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
No data available
Flash Point (°C)
No data available
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.