Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

218744

Sigma-Aldrich

Caspase-2 Inhibitor I

The Caspase-2 Inhibitor I controls the biological activity of Caspase-2. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Caspase-2 Inhibitor I, Z-VD(OMe)VAD(OMe)-FMK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H46FN5O11
Molecular Weight:
695.73
UNSPSC Code:
12352200
NACRES:
NA.54

Quality Level

Assay

≥95% (HPLC)
≥95% (TLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable and irreversible inhibitor of caspase-2 (ICH-1)
An irreversible inhibitor of caspase-2 (ICH-1).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
caspase-2
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Val-Asp(OMe)-Val-Ala-Asp(OMe)-CH₂F*

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Gregoli, P.A., and Bondurant, M.C. 1999. J. Cell Physiol. 178, 133.
Thornberry, N.A., and Lazebnik, Y. 1998. Science 281, 1312.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zahara L Chaudhry et al.
Brain sciences, 12(4) (2022-04-22)
The outbreak of COVID-19 caused by severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) signifies a serious worldwide concern to public health. Both transcriptome and proteome of SARS-CoV-2-infected cells synergize the progression of infection in host, which may exacerbate symptoms and/or
Zahara L Chaudhry et al.
Brain sciences, 10(11) (2020-11-05)
The current coronavirus pandemic caused by the severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) has resulted in a serious global health crisis. It is a major concern for individuals living with chronic disorders such as Parkinson's disease (PD). Increasing evidence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service