Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCP0063

Sigma-Aldrich

Calpastatin Peptide

≥95% (HPLC), Ac 184-210

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C142H230N36O44S1
Molecular Weight:
3177.63
UNSPSC Code:
12352202
NACRES:
NA.32

product name

Calpastatin Peptide Ac 184-210, ≥95% (HPLC)

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ac-Asp-Pro-Met-Ser-Ser-Thr-Tyr-Ile-Glu-Glu-Leu-Gly-Lys-Arg-Glu-Val-Thr-Ile-Pro-Pro-Lys-Tyr-Arg-Glu-Leu-Leu-Ala-NH2

Application

Calpastatin Peptide Ac 184-210 has been used as the endogenous calpain inhibitor to study its effects on myristoylated alanine-rich C kinase substrate (MARCKS) cleavage in mouse kidney cells. It has also been used as a calpain inhibitor to examine its effects on the formation of DNA-platinum adducts by cisplatin (CDDP) in neuronal cells.

Biochem/physiol Actions

Calpastatin, a component of the calpain/calpastatin system, is a calpain (calcium-dependent cysteine protease) inhibitor. The calpain/calpastatin system is involved in numerous membrane fusion events, such as neural vesicle exocytosis and platelet and red-cell aggregation.
Calpastatin, located in the cytosol, is involved in selective protein cleavage in response to calcium signaling.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calpain in the cleavage of alpha-synuclein and the pathogenesis of Parkinson's disease
Shams R, et al.
Progress in Molecular Biology and Translational Science, 167, 107-124 (2019)
Darrice S Montgomery et al.
American journal of physiology. Cell physiology, 313(1), C42-C53 (2017-05-05)
We previously demonstrated a role for the myristoylated alanine-rich C kinase substrate (MARCKS) to serve as an adaptor protein in the anionic phospholipid phosphate-dependent regulation of the epithelial sodium channel (ENaC). Both MARCKS and ENaC are regulated by proteolysis. Calpains
Aysel Cetinkaya-Fisgin et al.
Scientific reports, 10(1), 21889-21889 (2020-12-16)
Cisplatin is a commonly used chemotherapy agent with significant dose-limiting neurotoxicity resulting in peripheral neuropathy. Although it is postulated that formation of DNA-platinum adducts is responsible for both its cytotoxicity in cancer cells and side effects in neurons, downstream mechanisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service