Skip to Content
MilliporeSigma

208902

Calpastatin Peptide

A 27-amino acid, cell-permeable peptide encoded by exon 1B of human calpastatin that acts as a potent inhibitor of calpain I and calpain II (IC50 = 20 nM for purified rabbit calpain II).

Synonym(s):

Calpastatin Peptide, CS Peptide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C142H230N36O44S
Molecular Weight:
3177.63
NACRES:
NA.77
UNSPSC Code:
12352202
Assay:
≥95% (HPLC)
Form:
lyophilized solid
Storage condition:
OK to freeze, desiccated (hygroscopic)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic)

color

white

solubility

1% acetic acid: 1 mg/mL, water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A 27-amino acid, cell-permeable peptide encoded by exon 1B of human calpastatin that acts as a potent inhibitor of calpain I and calpain II (IC50 = 20 nM for purified rabbit calpain II). Does not inhibit either papain or trypsin. Blocks the down-regulation of protein kinase Cε (PKCε) in rat pituitary GH4C1 cells stimulated by thyrotropin-releasing hormone (TRH).
A 27-amino acid, peptide encoded by exon 1B of human calpastatin that acts as a cell-permeable and potent inhibitor of calpain I and calpain II (IC50 = 20 nM for purified rabbit calpain II). Does not inhibit either papain or trypsin. Blocks the down-regulation of protein kinase Cε (PKCε) in rat pituitary GH4C1 cells stimulated by thyrotropin-releasing hormone (TRH).

Biochem/physiol Actions

Product does not compete with ATP.
Reversible: no
Target IC50: 20 nM against calpain II
Cell permeable: yes
Primary Target
calpain 2

Physical form

Supplied as a trifluoroacetate salt.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Ac-Asp-Pro-Met-Ser-Ser-Thr-Tyr-Ile-Glu-Glu-Leu-Gly-Lys-Arg-Glu-Val-Thr-Ile-Pro-Pro-Lys-Tyr-Arg-Glu-Leu-Leu-Ala-NH₂
Eto, A., et al. 1995. J. Biol. Chem. 270, 25115.
Kawasaki, J., et al. 1989. J. Biochem. 106, 274.
Maki, M., et al. 1989. J. Biol. Chem. 264, 18866.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Explore all of our products under Calpastatin Peptide


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
208902-500UG04055977219500