Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

208726

Sigma-Aldrich

Calpain Inhibitor V

The Calpain Inhibitor V controls the biological activity of Calpain. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Calpain Inhibitor V, Mu-Val-HPh-FMK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30FN3O4
Molecular Weight:
407.48
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

orange-yellow

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A potent, cell-permeable, and irreversible inhibitor of calpain.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
calpain
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Mu-Val-HPh-CH₂F (Mu = morpholinoureidyl; HPh = homophenylalanyl)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Analysis Note

single spot by TLC

Other Notes

Esser, R.E., et al. 1994. Arthritis Rheum. 37, 236.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service