Skip to Content
MilliporeSigma
All Photos(12)

Documents

SAB4504446

Sigma-Aldrich

Anti-phospho-JAK1 (pTyr1022) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AIIDE, Anti-JAK1A, Anti-JAK1B, Anti-JTK3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 131 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr1022)

Gene Information

human ... JAK1(3716)

Immunogen

The antiserum was produced against synthesized peptide derived from human JAK1 around the phosphorylation site of Tyr1022.

Immunogen Range: 988-1037

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mari Ikai et al.
Veterinary dermatology, 34(4), 318-326 (2023-03-18)
Canine atopic dermatitis (cAD) is a disease associated with Type 2 helper T (Th2) immune responses in the acute phase of the disease. In humans, keratinocytes are activated by Th2 cytokines via the Janus kinase (JAK)/signal transducer and activator of
Takuwa Yasuda et al.
The Journal of clinical investigation, 126(6), 2064-2076 (2016-04-26)
Skin homeostasis is maintained by the continuous proliferation and differentiation of epidermal cells. The skin forms a strong but flexible barrier against microorganisms as well as physical and chemical insults; however, the physiological mechanisms that maintain this barrier are not
Hongnan Jiang et al.
Oncology reports, 48(5) (2022-10-01)
The present study aimed to investigate the underlying regulatory mechanism of MYCL proto‑oncogene (MYCL) in triple‑negative breast cancer (TNBC) progression. In vitro experiments were performed to confirm the functional roles of MYCL in TNBC, and its effects on the JAK/STAT3 pathway

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service