Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4300123

Sigma-Aldrich

Anti-phospho-JAK1 (pTyr1022) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-JAK1A antibody produced in rabbit, Anti-JAK1B antibody produced in rabbit, Anti-JTK3 antibody produced in rabbit, Anti-Janus kinase 1 (a protein tyrosine kinase) antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~130 kDa

species reactivity

rat, mouse, human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(K-E-YP-Y-T)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr1022)

Gene Information

human ... JAK1(3716)

General description

Janus kinase 1 (JAK1) is encoded by the gene mapped to human chromosome 1p31.3. The encoded protein is one of the four members of the mammalian JAK families. Members of JAK family contain an N-terminal FERM (four.1, ezrin, radixin, moesin) domain, through which they interact with cytokine receptors. The N-terminal end also contains an SH2-like domain. In addition, it also contains a kinase domain and a pseudo-kinase domain, which is catalytically inactive. JAK1 is ubiquitously expressed. When expressed without their receptors, JAKs reside in the cytosol. However, under normal cell conditions, they localize to plasma membrane and endosomes.

Immunogen

Peptide sequence around phosphorylation site of tyrosine 1022 (K-E-Y(p)-Y-T), according to the protein JAK1.

Application

Anti-phospho-JAK1 (pTyr1022) antibody produced in rabbit has been used in Western blot analysis.

Biochem/physiol Actions

Janus kinases (JAKs) are activated when cytokines bind with their receptors, upon which they phosphorylate the receptors and produce docking sites for signaling molecules. One of the main signaling molecules includes signal transducer and activator of transcription (Stat) family. JAK1 is needed by multiple cytokines, including the family using the shared receptor subunit called common γ chain (γc), such as interleukin (IL)-2, IL-4, IL-7, IL-9, IL-15 and IL-21. JAK1 KO (knockout) mice have early postnatal lethal or perinatal lethal phenotype due to neurological defects. In vitro studies show that JAK1S703I mutation functions as an activator of JAK-STAT pathways even in the absence of cytokines, and drives cellular proliferation. JAK1S703I mutant patient-derived xenograft (PDX) HCC (hepatocellular carcinoma) cells also show sensitivity to JAK1/2 inhibitor, ruxolitinib.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Tyrosine kinase of the non-receptor type, involved in the IFN-alpha/beta/gamma signal pathway. Kinase partner for the interleukin (IL)-2 receptor.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activating JAK1 mutation may predict the sensitivity of JAK-STAT inhibition in hepatocellular carcinoma.
Yang S, et al.
Oncotarget, 7(5), 5461?5469-5461?5469 (2016)
MiR-1246 Promotes LPS-Induced Inflammatory Injury in Chondrogenic Cells ATDC5 by Targeting HNF4γ.
W Da-Peng, et al.
Cellular Physiology and Biochemistry, 43(5), 2010-2021 (2017)
The Janus kinases (Jaks).
Yamaoka K et al
Genome Biology, 5(12), 253-253 (2004)
Da-Peng Wu et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 43(5), 2010-2021 (2017-10-23)
Osteoarthritis (OA) is a common inflammatory joint disease. miRNAs are associated with OA and functionally implicated in the pathogenesis of the disease. In the present study, we investigated the role of miR-1246 in the lipopolysaccharide (LPS)-induced inflammatory injury of ATDC5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service