MilliporeSigma
  • Phosphorylation of Janus kinase 1 and signal transducer and activator of transcription 3 and 6 in keratinocytes of canine atopic dermatitis.
Phosphorylation of Janus kinase 1 and signal transducer and activator of transcription 3 and 6 in keratinocytes of canine atopic dermatitis.
Veterinary dermatology (2023-03-18)
Mari Ikai, Mami Murakami, Toshitaka Kanei, Ryota Asahina, Munetaka Iwata, Hiroaki Kamishina, Sadatoshi Maeda
ABSTRACT
Canine atopic dermatitis (cAD) is a disease associated with Type 2 helper T (Th2) immune responses in the acute phase of the disease. In humans, keratinocytes are activated by Th2 cytokines via the Janus kinase (JAK)/signal transducer and activator of transcription (STAT) pathway. However, the activation of keratinocytes by Th2 cytokines in cAD has not yet been demonstrated. To evaluate keratinocyte activation based on the phosphorylation (p) of JAK1, STAT3 and STAT6. Seven dogs with cAD and three healthy dogs. Immunohistochemical analysis was performed to detect pJAK1, pSTAT3 and pSTAT6 in keratinocytes in normal canine skin, and the skin of atopic dogs. In the latter group samples were collected from both primary and secondary lesions, and nonaffected skin. The percentage of pJAK1-positive keratinocytes was significantly higher in primary cAD lesions than in healthy skin (p < 0.05). No significant differences were observed in pSTAT3-positive keratinocytes among the groups. The percentage of pSTAT6-positive keratinocytes was significantly higher in primary and secondary lesions than in healthy skin (p < 0.05, respectively). The novel finding in this study was the activation of keratinocytes as demonstrated by the phosphorylation of JAK1/STATs in lesional and nonlesional cAD skin. These results suggest the potential of not only JAK1, but also of STAT6 as therapeutic targets for cAD.
Sigma-Aldrich
Anti-phospho-JAK1 (pTyr1022) antibody produced in rabbit, affinity isolated antibody