Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4503022

Sigma-Aldrich

Anti-C1S, C-Terminal antibody produced in rabbit

affinity isolated antibody

Synonym(s):

C1 esterase, C1S, Complement C1s subcomponent, Complement component 1 subcomponent s

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 76 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... C1S(716)

General description

Anti-C1S Antibody detects endogenous levels of total C1S protein.
The C1S (complement C1s) gene is mapped to human chromosome 12p13 and codes for a subcomponent of C1 complex of the classical pathway.

Immunogen

The antiserum was produced against synthesized peptide derived from human C1S.

Immunogen Range: 541-590

Biochem/physiol Actions

Activation of C1s (complement C1s) along with C1r triggers complement cascade. C1s is responsible for the cleavage of complement component C4 and C2, which is necessary for complete activation of complement system. Studies determined that C1s specifically acts on Val or Leu at P(3) and Ala or Gly residues at P(2) positions. The C1S gene is known to be associated with lupus nephritis, with low gene expression observed in mice.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elucidation of the substrate specificity of the C1s protease of the classical complement pathway.
Kerr FK
The Journal of Biological Chemistry, 280(47), 39510-39514 (2005)
Gene expression analysis detected a low expression level of C1s gene in ICR-derived glomerulonephritis (ICGN) mice
Tamura K
Nephron: Experimental Nephrology, 123(3-4), 34-45 (2013)
Identification of a catalytic exosite for complement component C4 on the serine protease domain of C1s.
Duncan RC
Journal of Immunology, 189(5), 2365-2373 (2012)
A Molecular Switch Governs the Interaction between the Human Complement Protease C1s and Its Substrate, Complement C4
Perry AJ
The Journal of Biological Chemistry, 288(22), 15821?15829-15821?15829 (2013)
Complement genetics, deficiencies, and disease associations
Mayilyan KR
Protein & cell, 3(7), 487?496-487?496 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service