Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAB1059

Sigma-Aldrich

Anti-Pigment Epithelium Derived Factor Antibody, clone 10F12.2

clone 10F12.2, Chemicon®, from mouse

Synonym(s):

pigment epithelium-derived factor, proliferation-inducing protein 35, serine (or cysteine) proteinase inhibitor, clade F (alpha-2

antiplasmin, pigment epithelium derived factor), member 1, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10F12.2, monoclonal

species reactivity

human

species reactivity (predicted by homology)

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SERPINF1(5176)

General description

Pigment epithelium-derived factor (PEDF) was first identified as a 50 kDa secreted protein in conditioned medium from cultured fetal human retinal pigment epithelium (RPE) cells (Tombran-Tink, 1991). More recently, PEDF was shown to have neurotrophic activity in the retina in vivo (Cayouette, 1999). PEDF is also a neurotrophic and neuroprotective factor in other systems, including cultured cerebellar granule cells, primary hippocampal neurons, spinal cord motor neurons and amphibian photoreceptors cultured in the absence of RPE cells (Karakousis, 2001). The binding of PEDF to retinoblastoma and cerebellar granule cells may be receptor-mediated. Recently, PEDF was shown to be a potent inhibitor of angiogenesis in endothelial cell cultures and in rat cornea and retina in vivo (Dawson, 1999; Stellmach, 2001).

Specificity

Recognizes Pigment-Epithelium Derived Factor (PEDF).

Immunogen

Recombinant human GST-PEDF fusion protein. The sequence corresponding to amino
acids 196-347 of human PEDF tagged with GST on the N-terminal was expressed in E. Coli. GST-PEDF is about 46 kDa on SDS-PAGE.

Application

Anti-Pigment Epithelium Derived Factor Antibody, clone 10F12.2 is an antibody against Pigment Epithelium Derived Factor for use in ELISA, IC, IH & WB.
Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins
Western Blot:
Recognizes a protein with a molecular weight of ~46 kDa when tested against immunogen. Western blot positive control is available (Catalog number AG672)

ELISA:
A previous lot of this antibody was used on ELISA.

Immunocytochemistry:
A previous lot of this antibody was used on Immunocytochemistry.

Immunohistochemistry:
A previous lot of this antibody was used on frozen sections.

Optimal working dilutions must be determined by end user.

Quality

Routinely evaluated by Western Blot on human placenta lysate.

Western Blot Analysis:
1:1000 dilution of this lot detected pigment epithelium on 10 μg of human placenta lysate.

Target description

~46 kDa

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal IgG2a in buffer containing 0.02 M PBS, pH 7.6, 0.25 M NaCl with 0.1% sodium azide.

Storage and Stability

Stable for 6 months at 2-8°C in undiluted aliquots from date of receipt.

Analysis Note

Control
Eye tissue.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shou-Chuan Shih et al.
PloS one, 11(7), e0157647-e0157647 (2016-07-08)
Fibrogenesis is induced by repeated injury to the liver and reactive regeneration and leads eventually to liver cirrhosis. Pigment epithelium derived factor (PEDF) has been shown to prevent liver fibrosis induced by carbon tetrachloride (CCl4). A 44 amino acid domain
Mao Huang et al.
Neurotherapeutics : the journal of the American Society for Experimental NeuroTherapeutics, 15(3), 728-741 (2018-05-08)
Alzheimer's disease (AD) is the most common cause of dementia. Pigment epithelium-derived factor (PEDF), a unique neurotrophic protein, decreases with aging. Previous reports have conflicted regarding whether the PEDF concentration is altered in AD patients. In addition, the effect of
Célia Parinot et al.
International journal of molecular sciences, 25(12) (2024-06-27)
Among the myriad of existing tyrosine kinase receptors, the TAM family-abbreviated from Tyro3, Axl, and Mer tyrosine kinase (MerTK)-has been extensively studied with an outstanding contribution from the team of Prof. Greg Lemke. MerTK activity is implicated in a wide
Terakeith Lertsburapa et al.
Journal of neuroscience research, 75(5), 624-631 (2004-03-03)
Pigment epithelium-derived factor (PEDF) is a multifunctional protein with known anti-angiogenic and trophic properties, capable of promoting the survival and growth of Schwann cells (SC). Normal rat SCs and ganglioneuroma-derived human SCs secrete PEDF. The ability of normal SC to
Daisy S Nakamura et al.
BMC musculoskeletal disorders, 18(1), 39-39 (2017-01-27)
Inflammation is a major cause of cartilage destruction and leads to the imbalance of metabolic activities in the arthritic joint. Pigment epithelium-derived factor (PEDF) has been reported to have both pro- and anti-inflammatory activities in various cell types and to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service