Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4500012

Sigma-Aldrich

Anti-HDAC6 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

HD6, HDA6, Histone deacetylase 6, JM21

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 131 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HDAC6(10013)

General description

Histone deacetylase 6 (HDAC6) is a class IIb histone deacetylase which is localized in the cytosol. It has two catalytic domains. The protein is expressed in the heart, brain, colon, breast, ovary, pancreas and prostate glands. The gene encoding HDAC6 is localized on human chromosome Xp11.23. Anti-HDAC6 Antibody detects endogenous levels of total HDAC6 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human HDAC6.

Immunogen Range: 1166-1215

Biochem/physiol Actions

Histone deacetylase 6 (HDAC6) deacetylates substrates like α-tubulin. It is involved in endocytosis and autophagy. HDAC6 functions in cellular mechanisms connected to cancer and modulates cell motility.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen Hu et al.
Cell death & disease, 11(5), 328-328 (2020-05-10)
Ubiquitin-specific peptidase 10 (USP10) stabilizes both tumor suppressors and oncogenes in a context-dependent manner. However, the nature of USP10's role in non-small cell lung cancer (NSCLC) remains unclear. By analyzing The Cancer Genome Atlas (TCGA) database, we have shown that
Down-regulation of deacetylase HDAC6 inhibits the melanoma cell line A375.S2 growth through ROS-dependent mitochondrial pathway.
Bai J
PLoS ONE, 10(3), e0121247-e0121247 (2015)
Histone deacetylase 6 inhibition enhances oncolytic viral replication in glioma.
Nakashima H
The Journal of Clinical Investigation, 125(11), 4269-4280 (2015)
HDAC6 activity is a non-oncogene addiction hub for inflammatory breast cancers.
Putcha P
Breast Cancer Research, 17(1), 149-149 (2015)
Linlin Zhang et al.
Cancer biology & therapy, 15(11), 1561-1570 (2014-12-09)
Neuroblastoma is one of the most prevalent pediatric extracranial solid tumors and is often diagnosed after dissemination has occurred. Despite recent advances in multimodal therapies of this malignancy, its therapeutic efficacy remains poor. Novel treatment strategies are thus in great

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service