Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

AV31451

Sigma-Aldrich

Anti-HDAC6 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Histone deacetylase 6

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

pig, human, dog, rabbit, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

ChIP: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HDAC6(10013)

General description

HDAC6 is a histone deacetylase that associates with polyubiquitinated protein and also functions as a tubulin deacetylase. Additionally, HDAC6 is known to rescue neurodegeneration. It can also function as a mechanistic link between autophagy and ubiquitin-proteasome system during protein degradation.
Rabbit Anti-HDAC6 antibody recognizes canine, human, rat, bovine, and mouse HDAC6.

Immunogen

Synthetic peptide directed towards the N terminal region of human HDAC6

Application

Rabbit Anti-HDAC6 antibody can be used for western blot applications at 1.0μg/ml.

Biochem/physiol Actions

Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA. The protein belongs to class II of the histone deacetylase/acuc/apha family. It contains an internal duplication of two catalytic domains which appear to function independently of each other. This protein possesses histone deacetylase activity and represses transcription.

Sequence

Synthetic peptide located within the following region: VGLQGMDLNLEAEALAGTGLVLDEQLNEFHCLWDDSFPEGPERLHAIKEQ

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Udai Bhan Pandey et al.
Nature, 447(7146), 859-863 (2007-06-15)
A prominent feature of late-onset neurodegenerative diseases is accumulation of misfolded protein in vulnerable neurons. When levels of misfolded protein overwhelm degradative pathways, the result is cellular toxicity and neurodegeneration. Cellular mechanisms for degrading misfolded protein include the ubiquitin-proteasome system
Charlotte Hubbert et al.
Nature, 417(6887), 455-458 (2002-05-25)
Reversible acetylation of alpha-tubulin has been implicated in regulating microtubule stability and function. The distribution of acetylated alpha-tubulin is tightly controlled and stereotypic. Acetylated alpha-tubulin is most abundant in stable microtubules but is absent from dynamic cellular structures such as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service