Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC178

Sigma-Aldrich

Anti-P-Rex1 Antibody, clone 6F12

clone 6F12, from mouse

Synonym(s):

Phosphatidylinositol 3,4,5-trisphosphate-dependent Rac exchanger 1 protein, P-Rex1, PtdIns(3,4,5)-dependent Rac exchanger 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6F12, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PREX1(57580)

General description

Phosphatidylinositol 3,4,5-trisphosphate-dependent Rac exchanger 1protein (P-Rex1) is a guanine nucleotide exchange factor, which is predominantly expressed in the brain and leucocytes. It exchanges GDP for GTP in Rac 1, resulting in activation of Rac and its downstream signaling molecules that mediate such cellular processes as axon migration and differentiation.

Immunogen

Linear peptide corresponding to human P-Rex 1.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-P-Rex1, clone 6F12 is validated for use in IP, WB, IH for the detection of P-Rex1.
Western Blot Analysis: A representative lot was used by an independent laboratory to detect P-Rex 1 in PC3 cell lysate (Donald, S.,et al., 2010.), bone-marrow-derived neutrophils from mice (Qin, J., et al., 2009.), and mouse brain sections. (Welch, H, et al., 2005.)

Quality

Evaluated by Western Blot in THP-1 cell lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected P-Rex 1 in 10 µg of THP-1 cell lysate.

Target description

~186 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
THP-1 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emanuela Dylgjeri et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 25(18), 5623-5637 (2019-07-04)
DNA-dependent protein kinase catalytic subunit (DNA-PK) is a pleiotropic kinase involved in DNA repair and transcriptional regulation. DNA-PK is deregulated in selected cancer types and is strongly associated with poor outcome. The underlying mechanisms by which DNA-PK promotes aggressive tumor
Martin J Baker et al.
Cancers, 12(2) (2020-02-26)
The GTPase Rac1 is a well-established master regulator of cell motility and invasiveness contributing to cancer metastasis. Dysregulation of the Rac1 signaling pathway, resulting in elevated motile and invasive potential, has been reported in multiple cancers. However, there are limited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service