Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4300090

Sigma-Aldrich

Anti-phospho-MAPT (pSer262) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DDPAC antibody produced in rabbit, Anti-FLJ31424 antibody produced in rabbit, Anti-FTDP-17 antibody produced in rabbit, Anti-MAPTL antibody produced in rabbit, Anti-microtubule-associated protein tau antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~48 kDa
~62 kDa
~78 kDa

species reactivity

rat, human, mouse

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(I-G-SP-T-E)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer262)

Gene Information

human ... MAPT(4137)

General description

MAPT (microtubule associated protein tau) is located on human chromosome 17q21.3 It is highly expressed in neurons but is most prominent in axons

Immunogen

Peptide sequence around phosphorylation site of serine 262 (I-G-S(p)-T-E), according to the protein MAPT.

Biochem/physiol Actions

Removal of MAPT (microtubule associated protein tau) results in developmental delay and learning disability. It participates in the pathology of Alzheimer′s disease (AD). It helps in the assembly and maintenance of microtubule structure

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Promotes microtubule assembly and stability, and might be involved in the establishment and maintenance of neuronal polarity. The C-terminus binds axonal microtubules while the N-terminus binds neural plasma membrane components, suggesting that tau functions as a linker protein between both. Axonal polarity is predetermined by tau localization (in the neuronal cell) in the domain of the cell body defined by the centrosome. The short isoforms allow plasticity of the cytoskeleton whereas the longer isoforms may preferentially play a role in its stabilization.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linkage Disequilibrium and Association of MAPT H1 in Parkinson Disease
Skipper L
American Journal of Human Genetics, 75, 669-677 (2004)
The H1c haplotype at the MAPT locus is associated with Alzheimer's disease
Myers AJ
Human Molecular Genetics, 14, 2399-2404 (2005)
Microdeletion encompassing MAPT at chromosome 17q21.3 is associated with developmental delay and learning disability
Shaw-Smith C
Nature Genetics, 38(9), 1032-1037 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service