Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05-860

Sigma-Aldrich

Anti-Vitronectin Antibody, clone GMA-900

clone GMA-900, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

GMA-900, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... VTN(7448)

Specificity

Vitronectin

Immunogen

Purified human vitronectin

Application

Detect Vitronectin using this Anti-Vitronectin Antibody, clone GMA-900 validated for use in ELISA & WB.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

routinely evaluated by immunoblot on victronectin

Target description

Mr 54kDa

Linkage

Replaces: 04-1108

Physical form

DEAE purified
Format: Purified

Storage and Stability

stable 2 years at -20°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Wechsler-Reya
Trends in neurosciences, 24(12), 680-682 (2001-11-24)
Cerebellar granule cells are the most abundant neurons in the brain and are crucial to the circuitry that controls motor coordination. The proliferation of granule cell precursors (GCPs) is controlled by the secreted signaling molecule Sonic hedgehog (Shh), but the
I Schvartz et al.
The international journal of biochemistry & cell biology, 31(5), 539-544 (1999-07-10)
Vitronectin is a multifunctional glycoprotein present in blood and in the extracellular matrix. It binds glycosaminoglycans, collagen, plasminogen and the urokinase-receptor, and also stabilizes the inhibitory conformation of plasminogen activation inhibitor-1. By its localization in the extracellular matrix and its
Rui Chen et al.
Cold Spring Harbor protocols, 2015(7), 634-641 (2015-03-13)
Multiple platforms are available for whole-exome enrichment and sequencing (WES). This protocol is based on the Roche NimbleGen SeqCap EZ Exome Library SR platform, which enriches for ∼44 Mb of the human exonic regions. The SeqCap system uses 55- to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service