Skip to Content
MilliporeSigma
All Photos(1)

Documents

S8320

Supelco

SigmaMarker

high range, mol wt 36,000-200,000 Da

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

form

lyophilized powder

usage

sufficient for 10-20 large gels (16 × 18 cm)
sufficient for 20-30 mini-gels (10 × 10 cm)
1 vial sufficient for 50-100 PhastGels

mol wt

36,000-200,000 Da

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

SigmaMarkers are designed for use with Laemmli SDS-PAGE, the PhastSystem (Pharmacia) and Schagger and von Jagow SDS-PAGE (Tricine). SigmaMarkers cover the range of molecular weights common to most proteins or their subunits. The wide, high and low range markers are lyophilized with sample buffer so that they are ready-to-use after reconstitution with deionized water. The markers are formulated to yield a distribution of well-defined bands of approximately equal intensity after electrophoresis and Coomassie blue staining.

Other Notes

Protein (mol. wt.)
Glyceraldehyde-3-phosphate dehydrogenase, rabbit muscle (36,000)
Ovalbumin, chicken egg (45,000)
Glutamic dehydrogenase, bovine liver (55,000)
Albumin, bovine serum (66,000)
Phosphorylase B, rabbit muscle (97,000)
β-Galactosidase, E. coli (116,000)
Myosin, porcine heart (200,000)
Will yield bands for the following seven proteins from the SigmaMarker list with Coomassie Blue staining.

Reconstitution

Reconstitution with 100 μl of deionized water results in a solution containing 2-3.5 mg of protein per ml of 62 mM Tris-HCl, pH 6.8, 1 mM EDTA, 4% sucrose, 10 mM TCEP, 2% SDS, and 0.01% bromophenol blue.

Legal Information

SigmaMarker is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service