Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S7817

Sigma-Aldrich

Scriptaid

≥95%, solid

Synonym(s):

6-(1,3-Dioxo-1H, 3H-benzo[de]isoquinolin-2-yl)-hexanoic acid hydroxyamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18N2O4
CAS Number:
Molecular Weight:
326.35
MDL number:
UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥95%

form

solid

mp

160-161 °C

solubility

DMSO:methanol (1:1): complete 2 mg/ml, clear, colorless to faintly yellow
DMSO: 1 mg/mL
methanol: soluble

storage temp.

−20°C

SMILES string

ONC(=O)CCCCCN1C(=O)c2cccc3cccc(C1=O)c23

InChI

1S/C18H18N2O4/c21-15(19-24)10-2-1-3-11-20-17(22)13-8-4-6-12-7-5-9-14(16(12)13)18(20)23/h4-9,24H,1-3,10-11H2,(H,19,21)

InChI key

JTDYUFSDZATMKU-UHFFFAOYSA-N

Related Categories

General description

Scriptaid is a histone deacetylase (HDAC) inhibitor, which enhances global acetylation of histones. It improves transcriptional activity and protein expression. Scriptaid inhibits tumor growth. It decreases telomerase activity and stimulates glioma cell apoptosis.

Application

Scriptaid has been used in post activation of oocytes and embryo culture.
Scriptaid was used to enhance transcriptional activity in cloning procedures by somatic cell nuclear transfer.5,6

Biochem/physiol Actions

Histone deacetylase inhibitor with lower toxicity than trichostatin A.
Histone deacetylase inhibitor with lower toxicity than trichostatin A; used to enhance protein expression.
Scriptaid inhibits the cell cycle progression of ovarian cancer cells by inducing arrest in G0/G1 and/or G2/M phase. Treatment of cells with scriptaid results in loss of mitochondrial membrane potential and increased acetylation of H3 and H4 histone tails.2 Sciptaid induces expression of γ-globin in human erythroid progenitors via p38 signaling and may be a treatment option for sickle cell disease.3 It enhances the transcriptional activity and protein expression in mouse embryos. This is useful in producing cloned, inbred mouse embryos that develop normally into adulthood with regular reproductive ability.4

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Legal Information

Sold under license of U.S. Patent No. 6,544,957.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weihua Xu et al.
PloS one, 8(5), e64705-e64705 (2013-06-07)
Somatic cell nuclear transfer (SCNT) in mammalian cloning currently remains inefficient. Incomplete or erroneous epigenetic reprogramming of specialized donor somatic nuclear and resulting aberrant gene expression during development of cloned embryos is commonly believed as the main reason that causes
A novel histone deacetylase inhibitor, scriptaid, enhances expression of functional estrogen receptor ?(ER) in ER negative human breast cancer cells in combination with 5-aza 2?-deoxycytidine
Keen JC, et al.
Breast Cancer Research and Treatment, 81(3), 177-186 (2003)
Effects of combined treatment of MG132 and scriptaid on early and term development of porcine somatic cell nuclear transfer embryos
Mao J, et al.
Cellular Reprogramming, 14(5), 385-389 (2012)
Kiho Lee et al.
Molecular reproduction and development, 80(2), 145-154 (2012-12-15)
In general, pig embryos established by somatic cell nuclear transfer (SCNT) are transferred at the one-cell stage because of suboptimal embryo culture conditions. Improvements in embryo culture can increase the practical application of late embryo transfer. The goal of this
Nguyen Van Thuan et al.
Reproduction (Cambridge, England), 138(2), 309-317 (2009-05-13)
Since the birth of Cumulina, the first mouse clone produced by somatic cell nuclear transfer (SCNT), the success rate of cloning in mice has been extremely low compared with other species and most of the inbred mouse strains have never

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service