Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EPI008

Sigma-Aldrich

Histone Deacetylase (HDAC) Inhibitor Set I

Set includes 6 inhibitors

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.41

shipped in

wet ice

storage temp.

−20°C

General description

A convenient set of six selected individual HDAC (Histone Deacetylase) inhibitors.
  • Apicidin
CAS Number: 183506-66-3
Molecular Formula: C34H49N5O6
Molecular Weight: 623.8
  • M344
CAS Number: 251456-60-7
Molecular Formula: C16H25N3O3
Molecular Weight: 307.39
  • Sodium 4-phenylbutuyrate
CAS Number: 1716-12-7
Molecular Formula: C10H11O2·Na
Molecular Weight: 186.2
  • Splitomycin
CAS Number: 5690-03-9
Molecular Formula: C13H10O2
Molecular Weight: 198.2
  • Trichostatin A
CAS Number: 58880-19-6
Molecular Formula: C17H22N2O3
Molecular Weight: 302.4
  • Valproic Acid, Sodium Salt
CAS Number: 1069-66-5
Molecular Formula: C8H15O2·Na
Molecular Weight: 166.19

Biochem/physiol Actions

  • Apicidin - A potent, cell permeable inhibitor of histone deacetylase (IC50 = 700 pM for parasitic histone deacetylase). Inhibits HeLa cell proliferation (IC50 = 50-100 nM) and induces the transcriptional activation of p21 (WAF1) in a reversible manner. Also prevents H-ras-induced invasive phenotype of MCF-10A cells possibly by down regulating MMP-2 in a reversible manner.
  • M344 - An amide analog of Trichostatin A that potently Inhibits histone deacetylases (IC50 = 40 nM for rat liver HDAC and IC50 = 100 nM for maize HDAC). Induces differentiation and inhibits proliferation (~2 μM) of murine erythroleukemia cells.
  • Sodium 4-phenylbutuyrate - Inhibitor of histone deacetylase (HDAC). Anti-neoplastic agent and transcriptional regulator. Also acts as an inducer of tumor cytostasis and differentiation.
  • Splitomycin - Potent inhibitor of yeast NAD+ dependent histone deacetylase Sir2p (IC50= 60 μM). Sensitizes mammalian cells to a variety of DNA-damaging agents by abrogating Sir2p activity on p53. Acts by either altering or blocking access to the acetylated histone binding pocket.
  • Trichostatin A - Potent, reversible inhibitor of histone deacetylase. Mediates the activation of O6- methylguanine-DNA methyltransferase (MGMT). May be involved in cell cycle progression of several cell types, induces cell growth arrest at both G1 and G2/M phases. In some cases induces apoptosis.
  • Valproic acid sodium salt - A cell-permeable histone deacetylase 1 (HDAC1) inhibitor (IC50 = 400 μM) that exhibits anticancer, anti-inflammatory and neuroprotective properties. Recently it has been reported that Valproic acid enables reprogramming of primary human fibroblasts with only two factors, Oct4 and Sox2, without the need for the oncogenes c-Myc or Klf4.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service