Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P3201

Sigma-Aldrich

Platelet-Derived Growth Factor-BB human

≥98% (HPLC and SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

PDGF-BB

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Product Name

Platelet-Derived Growth Factor-BB human, PDGF-BB, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥98% (HPLC and SDS-PAGE)

form

lyophilized powder

potency

1.0-3.0 ng/mL EC50

quality

endotoxin tested

mol wt

~25 kDa

packaging

pkg of 10 μg
pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

≤1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... PDGFB(5155)

Looking for similar products? Visit Product Comparison Guide

General description

Platelet-derived growth factor (PDGF) is expressed as homo and heterodimers : PDGF-AB FDGF-AA, PDGF-BB, PDGF-CC and PDGF-DD. The PDGF gene is highly conserved mapped to human chromosome 7p22.3. The active form of Pdgf exists as disulfide-linked dimers. It consists of cystine-knot-fold growth factor domain, which is highly conserved, except for the N-terminal prodomain, which shows variation in length among the isoforms.

Application

Platelet-Derived Growth Factor-BB human has been used :
  • As a growth supplement in proximal tubule and glomerular explant cell culture
  • In three-dimensional scaffold preparation
  • To study the effect of brivanib on growth factor signaling pathways
  • In transwell migration assay

Biochem/physiol Actions

Platelet-derived growth factor (PDGF) elicits multifunctional actions with a variety of cells. It is mitogenic to mesoderm-derived cells, such as dermal and tendon fibroblasts, vascular smooth muscle cells, glial cells, and chondrocytes. PDGF significantly activates chemotaxis of neutrophils, monocytes, and fibroblasts. It increases the synthesis of phospholipids, cholesterol esters, glycogen and prostaglandins, and modulates LDL (low density lipoprotein) receptor binding.
PDGF stimulates fibroblast proliferation and extracellular matrix production in order to facilitate tissue repair. PDGF-BB isomer is found to be critical in angiogenesis and is highly expressed in tumor, mediating oncogenic pericyte recruitment. PDGF-BB is an important regulator of bone homeostasis, repair and regeneration.

Other Notes

Dimer of the B chain of human PDGF.

Physical form

Lyophilized from a 0.2 μm filtered solution in 30 mM acetic acid.

Analysis Note

The biological activity is measured by its ability to stimulate 3H-thymidine incorporation in quiescent NR6R-3T3 fibroblasts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasutoshi Takashima et al.
American journal of physiology. Renal physiology, 310(5), F395-F408 (2016-01-15)
Glomerular integrity and functions are maintained by growth factor signaling. Heparan sulfate, the major component of glomerular extracellular matrixes, modulates growth factor signaling, but its roles in glomerular homeostasis are unknown. We investigated the roles of heparan sulfate 6-O-endosulfatases, sulfatase
Clinical Findings Associated with a De Novo Partial Trisomy 10p11. 22p15. 3 and Monosomy 7p22. 3 Detected by Chromosomal Microarray Analysis.
Kohannim O,et al.
Case Reports in Genetics, 2011 , 1-6 (2011)
Matrix stiffness regulates migration of human lung fibroblasts.
Asano1 S, et al.
Physiological Reports, 5.9, 1-11 (2017)
Brivanib attenuates hepatic fibrosis in vivo and stellate cell activation in vitro by inhibition of FGF, VEGF and PDGF signaling.
Nakamura I, et al.
PLoS ONE, 9.4 (2014)
Rare incorporation of bone marrow-derived cells into kidney after folic acid-induced injury.
Szczypka MS, et al.
Stem Cells, 23.1, 44-54 (2005)

Articles

Platelet-Derived Growth Factor is the principal mitogen found in mammalian serum and is released from platelets during clot formation.

Protocols

TrueGel3D® Hydrogel Plate protocol guides high-throughput culture of human adipose MSCs for screening applications.

TrueGel3D® Hydrogel Plate protocol guides high-throughput culture of human adipose MSCs for screening applications.

TrueGel3D® Hydrogel Plate protocol guides high-throughput culture of human adipose MSCs for screening applications.

TrueGel3D® Hydrogel Plate protocol guides high-throughput culture of human adipose MSCs for screening applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service