Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S5376

Sigma-Aldrich

Methyl stearate

~99% (GC)

Synonym(s):

Methyl octadecanoate, Stearic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16CO2CH3
CAS Number:
Molecular Weight:
298.50
Beilstein:
1786213
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Assay

~99% (GC)

form

powder

bp

181-182 °C/4 mmHg (lit.)

mp

37-41 °C (lit.)

functional group

ester

lipid type

saturated FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OC

InChI

1S/C19H38O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h3-18H2,1-2H3

InChI key

HPEUJPJOZXNMSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl stearate or methyl octadecanoic acid is a saturated methyl ester and a representative key molecule of biodiesel fatty acid methyl esters (FAMEs) produced from vegetable oils. Methyl stearate occurs naturally in clove oil.

Application

Methyl stearate has been used as a reference standard
  • in gas chromatography (GC) to evaluate the absolute amount of methyl stearate among the fatty acid methyl esters of biodiesel
  • to evaluate the intramuscular fatty acid profile of fresh lamb meat in GC
  • to qualitatively analyse the transesterification reaction products by thin layer chromatography

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Incorporation of thyme leaves in the diet of pregnant and lactating ewes: Effect on the fatty acid profile of lamb
Nieto G, et al.
Small Ruminant Research, 105(1-3), 140-147 (2012)
Characterization and catalytic performance of potassium loaded on rice husk silica and zeolite nay for transesterification of jatropha seed oil
Montalbo KD, et al.
Quimica nova, 36(8), 1116-1120 (2013)
The effect of genotype and environment on biodiesel quality prepared from Indian mustard (Brassica juncea) grown in Australia
Wilkes MA, et al.
Industrial Crops and Products, 48, 124-132 (2013)
Volatile components of clove essential oil (Eugenia caryophyllus Spreng): neutral fraction
Muchalal M and Crouzet J
Agricultural and Biological Chemistry, 49(6), 1583-1589 (1985)
Effects of specific fatty acid methyl esters on diesel fuel lubricity
Geller DP and Goodrum JW
Fuel: The Science and Technology of Fuel and Energy, 83(17-18), 2351-2356 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service