Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
CH3(CH2)18COOCH3
CAS Number:
Molecular Weight:
326.56
UNSPSC Code:
12352211
NACRES:
NA.25
PubChem Substance ID:
EC Number:
214-304-8
Beilstein/REAXYS Number:
1791385
MDL number:
Quality Segment
assay
≥99% (capillary GC)
form
powder
bp
215-216 °C/10 mmHg (lit.)
mp
45-48 °C (lit.)
functional group
ester
lipid type
omega FAs
shipped in
ambient
storage temp.
−20°C
SMILES string
CCCCCCCCCCCCCCCCCCCC(=O)OC
InChI
1S/C21H42O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21(22)23-2/h3-20H2,1-2H3
InChI key
QGBRLVONZXHAKJ-UHFFFAOYSA-N
Still not finding the right product?
Explore all of our products under Methyl arachidate
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
235.4 °F - closed cup
flash_point_c
113.00 °C - closed cup
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.