Skip to Content
MilliporeSigma

1.09602

Methyl stearate

reference substance for gas-chromatography

Synonym(s):

Methyl stearate, Octadecanoic acid methyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H38O2
CAS Number:
Molecular Weight:
298.50
UNSPSC Code:
12352108
EC Index Number:
203-990-4
NACRES:
NA.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

form

solid

bp

215 °C/20 hPa

mp

37-39 °C

density

0.85 g/cm3 at 40 °C

storage temp.

no temp limit

SMILES string

O(C)C(=O)CCCCCCCCCCCCCCCCC

InChI

1S/C19H38O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h3-18H2,1-2H3

InChI key

HPEUJPJOZXNMSJ-UHFFFAOYSA-N

Analysis Note

Purity (GC): ≥ 99.0
Identity (IR-spectrum): conforms


Still not finding the right product?

Explore all of our products under Methyl stearate


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

230.0 °F - closed cup

flash_point_c

110.0 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
109602000504022536115511