Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

RAB0271

Sigma-Aldrich

Mouse IL-1 α ELISA Kit

for serum, plasma and cell culture supernatant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 10 pg/mL
standard curve range: 12.23-3000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Il1a(16175)

General description

The Mouse IL-1 α ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse IL-1 α in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse IL-1 α

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADDELISA 5X Assay/Sample Diluent Buffer D (Item K)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmed Al-Dwairi et al.
Experimental and therapeutic medicine, 16(2), 985-992 (2018-08-18)
Inflammatory bowel disease (IBD) is a chronic incurable condition characterized by relapsing inflammation of the gut. Intestinal smooth muscle cells (SMCs) are affected structurally and functionally during IBD due to excessive production of different inflammatory mediators. Metformin is a widely
Songfeng Zhao et al.
Chinese medical journal, 127(13), 2463-2469 (2014-07-06)
Epidermal burn injury may trigger significant apoptosis of the spleen cells, which might be caused by a burn-induced systemic inflammatory reaction. Heparin has been shown to possess anti-inflammatory properties. Interleukin 1 (IL-1) is centrally important among pro-inflammatory cytokines. We hypothesized
M Willi et al.
Clinical and experimental immunology, 178(1), 190-200 (2014-06-14)
Periodontitis is an inflammatory infectious disease that destroys the tooth-supporting tissues. It is caused by multi-species subgingival biofilms that colonize the tooth surface. Porphyromonas gingivalis, Treponema denticola and Tannerella forsythia (i.e. 'red complex' bacteria) are characteristic subgingival biofilm species. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service