MilliporeSigma
  • Metformin exerts anti-inflammatory effects on mouse colon smooth muscle cells in vitro.
Metformin exerts anti-inflammatory effects on mouse colon smooth muscle cells in vitro.
Experimental and therapeutic medicine (2018-08-18)
Ahmed Al-Dwairi, Mohammad Alqudah, Othman Al-Shboul, Mahmoud Alfaqih, Dana Alomari
ABSTRACT
Inflammatory bowel disease (IBD) is a chronic incurable condition characterized by relapsing inflammation of the gut. Intestinal smooth muscle cells (SMCs) are affected structurally and functionally during IBD due to excessive production of different inflammatory mediators. Metformin is a widely used antidiabetic agent known to exert several anti-inflammatory effects in different tissues independently from its hypoglycemic effect. The aim of the present study was to investigate the effect of metformin on expression and secretion of different cytokines and chemokines from mouse colon SMCs (CSMCs) following induction of inflammation with lipopolysaccharide (LPS) in vitro. CSMCs from male BALB/c mice were isolated and cultured in Dulbecco's modified Eagle's medium and treated with LPS (1 µg/ml) and 0, 5, 10 or 20 mM metformin for 24 h. Expression and secretion of tumor necrosis factor-α (TNF-α), interleukin-1α (IL-1α), macrophage colony stimulating factor (M-CSF), T cell activation gene-3 (TCA-3) and stromal cell-derived factor-1 (SDF-1) was evaluated by ELISA. LPS-treated CSMCs demonstrated significantly increased expression of TNF-α, IL-1α, M-CSF, TCA-3 and SDF-1 when compared with the control group (P<0.05). Co-treatment with metformin (5 and 10 mM) significantly reduced their expression by ~20-40% when compared with LPS treatment alone (P<0.05). Furthermore, secretion of TNF-α, IL-1α, M-CSF and TCA-3 into the conditioned media was significantly decreased by metformin (5 and 10 mM; P<0.05). In addition, metformin decreased levels of LPS-induced nuclear factor-κB phosphorylation. These data suggest that metformin may provide beneficial anti-inflammatory effects on CSMCs and it may be utilized as an adjunct therapy for patients suffering from IBD.
Sigma-Aldrich
Mouse Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse M-CSF / CSF1 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse SDF-1 α / CXCL12 α ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse TCA-3 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse IL-1 α ELISA Kit, for serum, plasma and cell culture supernatant