Skip to Content
MilliporeSigma

1.44253

Hycon® Agar Strips TC

Tryptic Soy Agar, Polysorbate 80, Lecithin, γ-irradiated, single packed of 50 units, suitable for air monitoring

Synonym(s):

Hycon Ready-to-use Agar Strips, TSA, Soybean Casein Digest, Agar Strips

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.74
UNSPSC Code:
41113035
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

EP, JP, USP

sterility

γ-irradiated

product line

RCS®

form

plate (ready-to-use)

feature

individually wrapped

composition

Lecithin , Polysorbate 80

packaging

single packed of 50 units

manufacturer/tradename

HYCON®

pH

7.3 ( in H2O)

application(s)

air monitoring, cosmetics
food and beverages
pharmaceutical

compatibility

for use with RCS® High Flow Touch

storage temp.

2-25°C

suitability

bacteria, fungi

General description

Agar Strips TC are ready-to-use culture media for assessment of airborne microorganisms RCS® Air Samplers. Each individual agar strip is welded into a re-sealable sleeve and 100% visually controlled, ensuring maximum safety in critical environments. Tryptic Soy Agar is filled in a flexible backing film. Tryptic Soy Agar (TSA, Tryptic Soy Agar Soybean Casein Digest Agar) is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The medium is supplemented with the neutralizers such as lecithin and polysorbate 80.

Application

Agar Strips are used for monitoring of ambient air and compressed gases with RCS® Air Samplers.

Legal Information

HYCON is a registered trademark of Merck KGaA, Darmstadt, Germany
RCS is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library