Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

R6258

Supelco

Packing reservoirs for non-jacketed LC columns

size 2,000 mL, column I.D. 4.8 cm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.52

technique(s)

LPLC: suitable

column I.D.

4.8 cm

size

2,000 mL

Looking for similar products? Visit Product Comparison Guide

General description

Supplied complete with all fittings necessary for connection to threaded columns. The top has 1/4 -28 fitting.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service