Skip to Content
MilliporeSigma
All Photos(1)

Documents

PZ0313

Sigma-Aldrich

PF-04802367

≥98% (HPLC)

Synonym(s):

N-(3-(1H-1,2,4-Triazol-1-yl)propyl)-5-(3-chloro-4-methoxyphenyl)oxazole-4-carboxamide, PF 04802367, PF-367, PF-4802367

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16ClN5O3
CAS Number:
Molecular Weight:
361.78
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

ClC1=CC(C2=C(C(NCCCN3N=CN=C3)=O)N=CO2)=CC=C1OC

InChI

1S/C16H16ClN5O3/c1-24-13-4-3-11(7-12(13)17)15-14(20-10-25-15)16(23)19-5-2-6-22-9-18-8-21-22/h3-4,7-10H,2,5-6H2,1H3,(H,19,23)

InChI key

RQFYFNAGNBUGFC-UHFFFAOYSA-N

Biochem/physiol Actions

PF-04802367 (PF-367) is a potent and specific ATP-competitive inhibitor of glycogen synthase kinase 3 (GSK-3α/β) with an IC50 value of 2.3 nM and >450-fold selectivity for GSK-3 in a panel of >400 neuroreceptors, enzymes and kinases. GSK-3 dysregulation has been implicated in the pathogenesis of neurological diseases including Alzheimer′s disease.
PF-04802367 is also known as [N-(3-(1H-1,2,4-triazol-1-yl)propyl)-5-(3-chloro-4-methoxyphenyl)oxazole-4-carboxamide) or PF-367. It regulates tau phosphorylation in the brain.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery of a Highly Selective Glycogen Synthase Kinase-3 Inhibitor (PF-04802367) That Modulates Tau Phosphorylation in the Brain: Translation for PET Neuroimaging
Liang S H, et al.
Angewandte Chemie (International Edition in English), 55(33), 9601-9605 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service