Skip to Content
MilliporeSigma

Skip To

SML2087

SJ000291942

≥98% (HPLC)

Synonym(s):

2-(4-Ethylphenoxy)-N-(4-fluoro-3-nitrophenyl)-acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C16H15FN2O4
CAS Number:
Molecular Weight:
318.30
UNSPSC Code:
12352202
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCC1=CC=C(OCC(NC2=CC([N+]([O-])=O)=C(F)C=C2)=O)C=C1

InChI

1S/C16H15FN2O4/c1-2-11-3-6-13(7-4-11)23-10-16(20)18-12-5-8-14(17)15(9-12)19(21)22/h3-9H,2,10H2,1H3,(H,18,20)

InChI key

YSZJLXPXVFGTNK-UHFFFAOYSA-N

Biochem/physiol Actions

Activator of Bone Morphogenetic Protein (BMP) signaling
SJ000291942 is an activator of Bone Morphogenetic Protein (BMP) signaling with an EC50 value < 1 μM. Bone Morphogenetic Proteins (BMPs) are a group of growth factors and cytokines were originally discovered by their ability to induce the formation of bone and cartilage, but are now considered to constitute a group of pivotal morphogenetic signals, orchestrating tissue architecture throughout the body. SJ000291942 was shown to ventralize zebrafish embryos, activate SMAD1/5/8 phosphorylation, and induce the differentiation of C2C12 myoblasts to osteoblasts in manner consistent with BMP4 activation.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1672SML2682SML2637
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

color

white to beige

color

white to beige

color

faint brown to very dark brown-red

color

white to beige


Storage Class

11 - Combustible Solids

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters