Skip to Content
MilliporeSigma
All Photos(2)

Documents

PHG0003

SAFC

MES hydrate

Pharma Manufacturing

Synonym(s):

2-(N-Morpholino)ethanesulfonic acid hydrate, 4-Morpholineethanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO4S · xH2O
CAS Number:
Molecular Weight:
195.24 (anhydrous basis)
MDL number:
UNSPSC Code:
12161700
NACRES:
NA.21

biological source

synthetic

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, microbial, and trace metals; tested

useful pH range

5.5-6.7

pKa 

6.1

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity, DNase, NICKase, RNase, and Protease; tested

SMILES string

O.OS(=O)(=O)CCN1CCOCC1

InChI

1S/C6H13NO4S.H2O/c8-12(9,10)6-3-7-1-4-11-5-2-7;/h1-6H2,(H,8,9,10);1H2

InChI key

MIIIXQJBDGSIKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Buffer quality is vital for the success of biopharmaceutical processes, because buffers are indispensable in nearly every production step.

Our broad portfolio of buffer materials manufactured under appropriate controls is tailored to your needs. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Application

MES is a biological buffer often referred to as a “Good′s” buffer. The pKa of MES is 5.96 which makes MES an ideal candidate for medias and protein based buffer formulations to maintain a stable environment in solution. MES is considered to be non-toxic to culture cell lines, highly water soluble and provides high-solution clarity.

MES is used in cell culture media, biopharmaceutical buffer formulations (both upstream and downstream), and diagnostic reagents. MES based buffers are used in purification bioprocesses of antibodies, peptides, proteins, blood components and growth factors.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tongtong Ma et al.
International journal of pharmaceutics, 473(1-2), 296-303 (2014-07-02)
Oral delivery of antigens is patient-friendly and efficient way of treating intestinal infections. However, the efficacy of oral vaccines is limited by degradation in the gastrointestinal (GI) tract and poor absorption by enterocytes and antigen-presenting cells (APC). Here we report
Tomoya Narawa et al.
Biological & pharmaceutical bulletin, 38(4), 545-551 (2015-04-04)
The stereoselective transport of methotrexate (L-amethopterin, L-MTX) and its enantiomer (D-amethopterin, D-MTX) by the rat proton-coupled folate transporter (rPCFT) were examined using rPCFT-expressing HEK293 cells. The initial rate of uptake of [3H]-L-MTX by the rPCFT followed Michaelis-Menten kinetics, with a
M Mirazul Islam et al.
Acta biomaterialia, 12, 70-80 (2014-12-03)
The implant-host interface is a critical element in guiding tissue or organ regeneration. We previously developed hydrogels comprising interpenetrating networks of recombinant human collagen type III and 2-methacryloyloxyethyl phosphorylcholine (RHCIII-MPC) as substitutes for the corneal extracellular matrix that promote endogenous
Xu Yu et al.
Biosensors & bioelectronics, 66, 520-526 (2014-12-17)
A novel highly sensitive colorimetric assay for DNA detection using cascade amplification strategy based on hybridization chain reaction and enzyme-induced metallization was established. The DNA modified superparamagnetic beads were demonstrated to capture and enrich the target DNA in the hybridization
Patricia Lopez-Sanchez et al.
Soft matter, 11(20), 4002-4010 (2015-04-23)
Cellulose hydrogels are extensively applied in many biotechnological fields and are also used as models for plant cell walls. We synthesised model cellulosic hydrogels containing hemicelluloses, as a biomimetic of plant cell walls, in order to study the role of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service