MilliporeSigma
  • M-cell targeted polymeric lipid nanoparticles containing a Toll-like receptor agonist to boost oral immunity.
M-cell targeted polymeric lipid nanoparticles containing a Toll-like receptor agonist to boost oral immunity.
International journal of pharmaceutics (2014-07-02)
Tongtong Ma, Lianyan Wang, Tingyuan Yang, Guanghui Ma, Siling Wang
ABSTRACT
Oral delivery of antigens is patient-friendly and efficient way of treating intestinal infections. However, the efficacy of oral vaccines is limited by degradation in the gastrointestinal (GI) tract and poor absorption by enterocytes and antigen-presenting cells (APC). Here we report ulex europaeus agglutinin-1 (UEA-1) conjugated poly (D,L-lactide-co-glycolide) (PLGA)-lipid nanoparticles (NP) containing a Toll-like receptor (TLR)-agonist monophosphoryl lipid A (MPL) as an oral vaccine delivery system. The uniform-sized PLGA-lipid NPs (simplified as lipid NPs) were produced by the premix membrane emulsification method. They can protect the entrapped model antigen ovalbumin (OVA) from exposure to the GI tract and release the OVA in a controlled manner. With UEA-1 and MPL modification, the UEA-MPL/lipid NPs can be effectively transported by M-cells and captured by mucosal dendritic cells (DCs). After in vivo vaccination, the OVA-UEA-MPL/lipid NPs stimulated the most effective mucosal IgA and serum IgG antibodies during the oral formulations. These results suggest that this MPL containing M-cell targeted lipid NP can potentially be used as a universally robust oral vaccine delivery system.
Product Description
SAFC
MES monohydrate
SAFC
MES hydrate
Sigma-Aldrich
MES hydrate, ≥99.5% (titration), pH 2.5-4.0 (0.5 M in H2O), BioXtra
Sigma-Aldrich
MES hydrate, ≥99.5% (titration)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
MES hydrate, BioPerformance Certified, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
MES monohydrate, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
MES hydrate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
MES monohydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Ethanesulfonic acid, 95%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)