Skip to Content
MilliporeSigma
All Photos(1)

Documents

69889

Sigma-Aldrich

MES monohydrate

BioUltra, for molecular biology, ≥99.5% (T)

Synonym(s):

2-Morpholinoethanesulfonic acid hydrate,, 2-morpholin-4-ylethanesulfonic acid;hydrate, 2-(N-Morpholino)ethanesulfonic acid, 4-Morpholineethanesulfonic acid monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H13NO4S · H2O
CAS Number:
Molecular Weight:
213.25
Beilstein:
6350956
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
32129211
PubChem Substance ID:
NACRES:
NA.25

grade

for molecular biology

Quality Level

product line

BioUltra

Assay

≥99.5% (T)

form

powder or crystals

impurities

DNases, none detected
Insoluble matter, passes filter test
Phosphatases, none detected
Proteases, none detected
RNases, none detected

ign. residue (900 °C)

≤0.05% (as SO4)

pH

2.5-4.0 (25 °C, 0.5 M in H2O)

useful pH range

5.5-6.7

pKa (25 °C)

6.1

mp

>300 °C (lit.)

solubility

H2O: 0.5 M at 20 °C, clear, colorless

density

10.66 g/mL

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.5 M in H2O

UV absorption

λ: 260 nm Amax: 0.025
λ: 280 nm Amax: 0.020

SMILES string

O.OS(=O)(=O)CCN1CCOCC1

suitability

suitable for DNase I test
suitable for Western blot
suitable for molecular biology

application(s)

clinical research
diagnostic assay manufacturing
life science and biopharma

foreign activity

DNase, none detected
NICKase, none detected
RNase, none detected
protease, none detected

InChI

1S/C6H13NO4S.H2O/c8-12(9,10)6-3-7-1-4-11-5-2-7;/h1-6H2,(H,8,9,10);1H2

InChI key

MIIIXQJBDGSIKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

MES Buffer, known as one of the "Good′s" buffers designed for biological applications, plays a pivotal role in various aspects of research. This zwitterionic, morpholinic buffer is particularly valuable for its buffering capabilities within a pH range of 5.5 to 6.7. MES monohydrate, finds extensive utility in maintaining a stable environment in solutions used for cell culture media, protein-based buffer formulations, and electrophoresis running buffers. Its significance extends to the purification processes of antibodies, peptides, proteins, blood components, and growth factors. Additionally, MES stands out as an excellent choice for capillary electrochromatography, offering low ionic mobility. It serves as a safer alternative to toxic agents like cacodylate and non-zwitterionic buffers such as citrate and malate. While MES excels in buffering, it′s important to note that it is not recommended for use at pH 7.4, and alternative buffers should be explored in such cases. MES Monohydrate is also used to fine-tune the pH of growth media, stabilize enzymatic solutions, and as a component of PAGE running buffer in various experimental settings.

Application

MES Monohydrate has been used:
  • In the preparation of a solution used for the suspension and dilution of protoplasts to measure the density and number of protoplasts using a haemocytometer
  • As a component of Murashige and Skoog basal salt media
  • In the preparation of total ionic strength adjustment buffer (TISAB)
  • as a constituent in mobile phase solutions employed for the analysis of high molecular weight species and charge-related variants of biopharmaceutical proteins through SEC, HIC, and IEX chromatographic techniques

Features and Benefits

  • Suitable as a Buffer component, for Electrophoresis and Molecular Biology
  • Effective Buffering from pH 2.5-4.0 (25 °C, 0.5 M in H2O) with a pKa of 6.1 (25 °C)
  • Free from DNase, NICKase, RNase, and Protease

Preparation Note

A buffer using MES free acid can be prepared by titrating the free acid with sodium hydroxide to the desired pH (pKa =±1). Alternatively, volumes of equimolar MES free acid and sodium MES can be mixed to attain the desired pH. Standard mixing tables using stock solutions to prepare a buffer of a given pH have been published.

Other Notes

Solubility/Stability: MES is soluble in water, giving a clear colourless solution at concentrations of 0.5 M or higher. The pH of a solution should be between 2.5 and 5, depending on the concentration. Solutions are stable at 2-8C for months.
Sterilization: Sterilization should be by filteration through 0.2 uM filters. Autoclaving is not recommended by any sulfonic acid buffers. If buffers must be nuclease-free, it is best to treat the water, and then add the buffer solids after autoclaving. When MES solutions are autoclaved, they turn yellow (although pH does not change measurably. The identity of the yellow breakdown product is unknown.
For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[14 C] Glucose Cell Wall Incorporation Assay for the Estimation of Cellulose Biosynthesis.
Brabham, CJ and Stork, J
Bio-protocol, 5 (18), 1-1 (2015)
In vivo monitoring of plant small GTPase activation using a Forster resonance energy transfer biosensor.
Wong, HL, et al.
Plant methods, 14 (1), 56-56 (2018)
Development of a tubular fluoride potentiometric detector for flow analysis: Evaluation and analytical applications.
Santos, J and R. A. S. Lapa
Analytica Chimica Acta, 583 (2), 429-436 (2007)
Good, NE, et al.
Biochemistry, 5, 467-477 (1966)
Dawson, R.M.C. et al.
Data for Biochemical Research, 410- 410 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service