Skip to Content
MilliporeSigma
All Photos(1)

Documents

P7032

Sigma-Aldrich

Pyruvate Dehydrogenase from porcine heart

buffered aqueous glycerol solution, 0.1-1.0 units/mg protein (Lowry)

Synonym(s):

PDH, Pig heart pyruvate dehydrogenase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

buffered aqueous glycerol solution

Quality Level

specific activity

0.1-1.0 units/mg protein (Lowry)

concentration

≥5.0 mg/mL

impurities

polyethylene glycol, trace

foreign activity

α-ketoglutarate dehydrogenase ≤15%

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Pyruvate dehydrogenase contributes to linking the glycolysis metabolic pathway to the citric acid cycle and releases energy via NADH. It is used to study how aerobic training effects pyruvate dehydrogenase . It is also used to study the interaction between pyruvate dehydrogenase kinase and the pyruvate dehydrogenase complex as well as the function and regulation of the pyruvate dehydrogenase complex. Product P7032 is provided in a buffered aqueous glycerol solution and is from porcine heart.
The enzyme has been used in a primary enzyme assay to test the ability of certain compounds to reduce PDK- (pyruvate dehydrogenase kinase) catalyzed PDC (pyruvate dehydrogenase complex) inactivation.

Biochem/physiol Actions

Pyruvate dehydrogenase is involved in the decarboxylation of pyruvate and reductive acetylation of lipoic acid within the pyruvate dehydrogenase complex (PDC). Pyruvate dehydrogenase kinase (PDK) inactivates pyruvate dehydrogenase while pyruvate dehydrogenase phosphatase activates pyruvate dehydrogenase.

Unit Definition

One unit will convert 1.0 μmole of β-NAD to β-NADH per min at pH 7.4 at 30 °C in the presence of saturating levels of Coenzyme A.

Physical form

Supplied as a 50% glycerol solution containing ~9 mg/mL bovine serum albumin, 30% sucrose, 1.5 mM EDTA, 1.5 mM EGTA, 1.5 mM 2-mercaptoethanol, 0.3 TRITON® X-100, 0.003% sodium azide, and 15 mM potassium phosphate, pH 6.8

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Insulin stimulation of pyruvate dehydrogenase in an isolated plasma membrane-mitochondrial mixture occurs by activation of pyruvate dehydrogenase phosphatase.
D A Popp et al.
The Journal of biological chemistry, 255(16), 7540-7543 (1980-08-25)
Function and regulation of mammalian pyruvate dehydrogenase complex. Acetylation, interlipoyl acetyl transfer, and migration of the pyruvate dehydrogenase component.
R L Cate et al.
The Journal of biological chemistry, 254(5), 1659-1665 (1979-03-10)
T D Aicher et al.
Bioorganic & medicinal chemistry letters, 9(15), 2223-2228 (1999-08-28)
Several oximes of triterpenes with a 17-beta hydroxyl and abietane derivatives are inhibitors of pyruvate dehydrogenase kinase (PDK) activity. The oxime 12 and dehydroabietyl amine 2 exhibit a blood glucose lowering effect in the diabetic ob/ob mouse after a single
Naoyuki Makita et al.
The Journal of pharmacy and pharmacology, 69(1), 43-51 (2016-10-21)
Recently, there has been a growing interest in the mechanism of action of dichloroacetate (DCA) for T-cell differentiation; however, this mechanism has not been elucidated in detail. Therefore, this study aimed to investigate the mechanism of action of DCA for
Suzanne D DeBrosse et al.
Molecular genetics and metabolism, 107(3), 394-402 (2012-10-02)
Pyruvate dehydrogenase complex (PDC) deficiency is a relatively common mitochondrial disorder that primarily presents with neurological manifestations and lactic acidemia. We analyzed the clinical outcomes and neurological features of 59 consented symptomatic subjects (27 M, 32 F), who were confirmed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service