Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

UFC9100

Millipore

Amicon® Ultra Centrifugal Filter, 100 kDa MWCO

greener alternative

sample volume 15 mL, regenerated cellulose membrane, MWCO 100 kDa

Synonym(s):

100 kDa MWCO, 15 mL sample volume, Centrifugal concentrator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
eCl@ss:
36100101
NACRES:
NB.24

material

polyethylene cap
polyethylene liner
polypropylene tube
regenerated cellulose membrane
regenerated cellulose support
styrene-butadiene housing

Quality Level

sterility

non-sterile

product line

Amicon®

packaging

pkg of 24 pkg
pkg of 8 pkg
pkg of 96 pkg

manufacturer/tradename

Amicon® Ultra

sustainability

Greener Alternative Product

parameter

15 mL sample volume

technique(s)

protein extraction: suitable
protein purification: suitable

L

12.1 cm (4.807 in.)

diam.

2.97 cm

filtration area

7.6 cm2

volume

15 mL

working volume

≤15 mL
15 mL

pore size

100 kDa MWCO

greener alternative category

shipped in

ambient

General description

Amicon® Ultra 15 mL centrifugal devices with Ultracel® 100,000 Dalton molecular weight cutoff filters provide fast ultrafiltration, and reliable retention of macromolecules. For example, IgG (156 kDa) at 1 mg/ml initial concentration is retained at >90% .

The vertical design and available membrane surface area provide fast sample processing, high sample recovery (typically greater than 90% of dilute starting solution), and the capability for 80-fold concentration. Solute polarization and subsequent fouling of the membrane are minimized by the vertical design, and a physical deadstop in the filter device prevents spinning to dryness and potential sample loss.

The concentrate is collected from the filter device sample reservoir using a pipetter, while the ultrafiltrate is collected in the provided centrifuge tube. The device can be spun in a swinging bucket or fixed angle rotor. With their vertical membrane design and deadstop, Amicon® Ultra 15 mL filters deliver the highest recovery with short spin times.
We are committed to bringing you Greener Alternative Products, which have been re-engineered to enable more sustainable research solutions. The inner filtration device is made from 60% bio-circular material allocated using the ISCC mass balance approach. Click here for more information.

Application

  • Ultrafiltration with 100,000 Dalton molecular weight cutoff Ultracel® membrane.
  • Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms.
  • Purification of macromolecular components found in tissue culture extracts or cell lysates and protein removal prior to HPLS.
  • Concentration of dilute or pre-purified proteins from column eluents.
  • Desalting, buffer exchange and protein dialysis.

Features and Benefits

  • Ultracel® regenerated cellulose membrane with 100 kDa NMWCO delivers retentate recovery of greater than 90%.
  • Vertical membrane reduces concentration polarization for ultra-fast spin times (as fast as 10–15 minutes).
  • Heat-sealed membrane minimizes downstream extractables.
  • 100% integrity tested for reliable performance.
  • Convenient sample monitoring with translucent housing and volume gradations.
  • Direct pipettor sample access eliminates processing step to recover concentrate.
  • High concentration factors of 80–100X.

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Filtration methods and membrane filter selection in nanoparticle purification and production.

Filtration methods and membrane filter selection in nanoparticle purification and production.

Filtration methods and membrane filter selection in nanoparticle purification and production.

Filtration methods and membrane filter selection in nanoparticle purification and production.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service