MilliporeSigma
  • A CD153+CD4+ T follicular cell population with cell-senescence features plays a crucial role in lupus pathogenesis via osteopontin production.
A CD153+CD4+ T follicular cell population with cell-senescence features plays a crucial role in lupus pathogenesis via osteopontin production.
Journal of immunology (Baltimore, Md. : 1950) (2015-05-15)
Suhail Tahir, Yuji Fukushima, Keiko Sakamoto, Kyosuke Sato, Harumi Fujita, Joe Inoue, Toshimitsu Uede, Yoko Hamazaki, Masakazu Hattori, Nagahiro Minato
ABSTRACT
Immune aging results in diminished adaptive immunity and increased risk for autoimmunity. We previously reported a unique PD-1(+) CD44(high)CD4(+) T cell population that increases with age in normal mice. In this study, we indicate that the age-dependent PD-1(+) CD44(high)CD4(+) T cells develop as unique T follicular (TF) cells in a B cell-dependent manner and consist of two subpopulations, as follows: CD153(+) cells preferentially secreting abundant osteopontin on TCR stimulation and CD153(-) cells that are apparently TCR anergic. These unique TF cells with essentially similar features increase much earlier and are accumulated in the spontaneous germinal centers (GCs) in lupus-prone female BWF1 (f-BWF1) mice. These TF cells showed characteristic cell-senescence features and developed in association with extensive CD4(+) T cell proliferation in vivo, suggesting replicative senescence. Although the CD153(+) TF cells were defective in proliferation capacity, they were quite stable and specifically responded to self GC-B cells to secret abundant osteopontin, which inhibited B cell receptor-induced GC-B cell apoptosis in f-BWF1 mice. Transfer of CD153(+) PD-1(+) CD4(+) T cells promoted the growth of spontaneous GCs, whereas administration of anti-osteopontin Ab suppressed GC enlargement and anti-nuclear Ab production and ameliorated clinical lupus nephritis of f-BWF1 mice. Current results suggest that senescent CD153(+) TF cells generated as a consequence of extensive endogenous CD4(+) T cell proliferation play an essential, if not sufficient, role in lupus pathogenesis in lupus-prone genetic background and may also contribute to an increased autoimmunity risk with age.
Sigma-Aldrich
Osteopontin human, recombinant, expressed in HEK 293 cells, ≥97% (SDS-PAGE), ≥97% (HPLC), suitable for cell culture
Sigma-Aldrich
Osteopontin from mouse, >95% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Osteopontin human, recombinant, expressed in NSO cells, ≥95% (SDS-PAGE), lyophilized powder, suitable for cell culture