Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O3514

Sigma-Aldrich

Bovine Osteopontin

from bovine milk, powder, suitable for cell culture

Synonym(s):

Bone Sialoprotein-1, Early T Lymphocyte Activation Protein-1 (ETA-1), OPN, Secreted Phosphoprotein 1 (Spp1)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

product name

Osteopontin from bovine milk, lyophilized powder

biological source

bovine milk

Quality Level

sterility

sterile

form

lyophilized powder

mol wt

~60 kDa

packaging

pkg of 50 μg

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

Osteopontin (OPN) is a multifunctional protein. It is released by macrophages, T cells and epithelial cells. It has 261 amino acid residues with a molecular mass of 29,283 Da. Osteopontin is also called as uropontin. Osteopontin from bovine milk is highly glycosylated and phosphorylated. SPP1 gene (secreted phosphoprotein 1) codes for osteopontin. This gene is located on human chromosome 4 and cow chromosome 6. It has seven exons and six introns.

Biochem/physiol Actions

A secreted, highly acidic, calcium binding, phosphorylated glycoprotein that is chemotactic for macrophages, smooth muscle cells, endothelial cells, and glial cells. In activated macrophages, OPN inhibits nitric oxide production and cytotoxicity. Human, mouse, rat, pig, and bovine OPN share 40% amino acid sequence identity.
Osteopontin stimulates cell mediated responses, chemotaxis of inflammatory cells and anti-inflammatory responses. It binds to av-containing integrins and stimulate cell adhesion and migration. Osteopontin promotes Th1 type immunity.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 2.5 mg bovine serum albumin.

Analysis Note

The biological activity is measured by its ability to mediate 293 cell adhesion.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guidebook to the Extracellular Matrix, Anchor, and Adhesion Proteins (1999)
Milk and Milk Products in Human Nutrition (2011)
Molecular analysis of the Bacillus subtilis bacteriophage SPP1 region encompassing genes 1 to 6: The products of gene 1 and gene 2 are required for pac cleavage.
Chai S, et al.
Journal of Molecular Biology, 224(1), 87-102 (1992)
D D Hu et al.
The Journal of biological chemistry, 270(44), 26232-26238 (1995-11-03)
Osteopontin (OPN) is an extracellular matrix protein that binds to integrin alpha v beta 3. Here we demonstrate that two other integrins, alpha v beta 1 and alpha v beta 5, are also receptors for OPN. Human embryonic kidney 293
Milk Proteins: From Expression to Food (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service