Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK537

Sigma-Aldrich

D-Amino Acid Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

application(s)

food and beverages

detection method

colorimetric
fluorometric

relevant disease(s)

genitourinary disorders

shipped in

wet ice

storage temp.

−20°C

General description

D-amino acids are not as widespread as their enantiomeric counterparts in proteins, but they can be found in organisms ranging from bacteria (cell walls and antibiotics) to mammals (central nervous systems). The presence of D-amino acids in food is also of considerable interest. Racemization of L-amino acids during food processing may affect food quality and nutritional value.

Features and Benefits

  • Linear detection range: 0.86 to 500 µM for the colorimetric assay and 0.18 to 50 µM for the fluorimetric assay.
  • Compatible with high-throughput handling systems.

Suitability

The kit is suitable for D-amino acid determination in tissue, milk, and other biological preparations.

Principle

The D-Amino Acid Assay Kit uses an enzyme-catalyzed oxidation of D-amino acids to convert a dye into a colored and fluorescent form. The absorbance at 570 nm or fluorescence intensity at λEx = 530 nm/λEm = 585 nm is directly proportional to the D-amino acid concentration in the sample.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service