Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK466

Sigma-Aldrich

Free Fatty Acid Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

application(s)

agriculture
food and beverages
pharmaceutical

detection method

colorimetric
fluorometric

relevant disease(s)

diabetes; pediatric diseases; endocrinological disorders; neurological disorders

shipped in

wet ice

storage temp.

−20°C

General description

Fatty acids are aliphatic monocarboxylic acids ubiquitously found in animal or vegetable fat, oil, and wax. Fatty acids play important roles in cellular synthesis, energy metabolism, and are implicated in diverse disorders such as diabetes mellitus, sudden infant death syndrome, and Reye Syndrome.

Features and Benefits

  • Linear detection range: colorimetric assay 7 - 1000 µM, fluorometric assay 7 - 100 µM fatty acid.
  • Compatible with high-throughput handling systems.

Suitability

The kit is suitable for detecting free fatty acids in biological samples such as serum, plasma, urine, saliva, milk, and cell cultures, and in food and agriculture products, as well as studying the effects of drugs on free fatty acid metabolism.

Principle

The Free Fatty Acid Assay Kit provides a simple, one-step and high-throughput assay for measuring Free Fatty acids. In this assay, free fatty acids are enzymatically converted to acyl-CoA and subsequently to H2O2. The resulting H2O2 reacts with a specific dye to form a pink colored product. The optical density at 570 nm or fluorescence intensity (λEx = 530 nm/λEm = 585 nm) is directly proportional to the free fatty acid concentration in the sample.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service