RIPA buffer as the lysis reagent is acceptable, provided that the concentration of the RIPA buffer is consistent with that of the control.
MAK171
Colorimetric Biotin Assay Kit
sufficient for 200 colorimetric assays
Synonym(s):
Biotin Quantitation Kit
About This Item
Recommended Products
detection method
colorimetric
relevant disease(s)
dermatological diseases; neurological disorders
shipped in
dry ice
storage temp.
−20°C
Related Categories
General description
Suitability
Principle
Preparation Note
Precautions and Disclaimer: This product is for R&D;use only, not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Is it possible to use RIPA buffer as the lysis reagent with the Colorimetric Biotin Assay Kit, product number MAK171-1KT, to extract proteins and biotin from samples such as brain parenchymal tissues?
1 answer-
Helpful?
-
-
What are the instructions for running the kit with catalog number MAK171? Additionally, needs more information to understand the purpose of this kit and how to perform calculations if purchased.
1 answer-
The Colorimetric Biotin Quantitation Kit with catalog number MAK171 offers a convenient method for estimating the molar ratio of biotin to protein in biotin-protein conjugates or for quantifying biotin concentration in a solution. The assay utilizes HABA [2-(4-Hydroxyphenylazo)benzoic acid], a reagent that exhibits significant spectral changes when bound to avidin. Biotin readily displaces HABA from the HABA/Avidin complex, leading to a decrease in absorption at 500 nm. The kit provides an optimal ratio of Avidin and HABA and is most effective for determining biotin concentration in the range from 2–16 mM. The assay can be carried out in a cuvette or microplate format.
Helpful?
-
-
Pourquoi la concentration du control positif n'est pas donnée ? Question de savoir si le test a bien fonctionné et la concentration de notre échantillon est juste.
1 answer-
The positive control is a vial of 100uL of 100uM d-biotin.
Helpful?
-
-
Can I use it to measure serum samples?
1 answer-
This product is suitable for biotin-containing samples. The range of detection for this kit is 2 - 16 uM. Biotin concentrations would need to be within this range. Please see the link below to review the product datasheet for this kit:
https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/116/046/mak171bul.pdfHelpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service