Skip to Content
MilliporeSigma
All Photos(1)

Documents

MAK137

Sigma-Aldrich

Phospholipase D Activity Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

PLD Activity Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric or fluorometric tests

detection method

colorimetric
fluorometric

relevant disease(s)

cancer

storage temp.

−20°C

General description

Phospholipase D (PLD) catalyzes the hydrolysis of the phosphodiester bond of glycerophospholipids to generate phosphatidic acid and choline. It is ubiquitously expressed and is implicated in immune function and cancer inhibition. Abnormalities in PLD expression have been associated with human cancers such as breast, renal, gastric and colorectal cancer.

Application

Phospholipase D (PLD) Activity Assay Kit has been used to measure PLD activity in the liver and lysosomal fractions of the mice liver.
Suitable for the detection of phospholipase D activity in biological samples. This kit can be used to study the effects of various compounds on phospholipase D activity.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the quantitative determination of phospholipase D activity and evaluation of drug effects on phospholipase D metabolism.

Principle

In this assay, PLD hydrolyzes phosphatidylcholine to choline, which is determined using choline oxidase resulting in a colorimetric (570 nm)/fluorometric (λex = 530/λem = 585 nm) product, proportional to the PLD activity in the sample. Unit definition: one unit of PLD catalyzes the formation of 1 mmole choline per minute under the assay conditions (pH 7.4).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phospholipase D.
Su W and Michael A F
Handbook of Cell Signaling, 1167-1176 (2009)
Walid El Kayal et al.
Horticulture research, 4, 17042-17042 (2017-11-09)
Raspberry (Rubus spp.) is an economically important crop with a restricted growing season and very limited fruit shelf-life due to its extreme tenderness. In order to prolong its shelf life, an aqueous composition containing hexanal as the key active ingredient
Anne Lise Ferrara et al.
Journal of immunology research, 2019, 5836476-5836476 (2019-07-30)
Mastocytosis is a condition characterized by the expansion and accumulation of mast cells (MCs) in various organs. The symptoms are related to the increased release of MC-derived mediators that exert local and distant effects. MCs are a source and target
Przemysław Bernat et al.
Journal of plant physiology, 229, 158-163 (2018-08-11)
2,4-dichlorophenoxyacetic acid (2,4-D) is among the most commonly used herbicides applied for weed control during wheat cultivation. However, its application could affect wheat growth. The present study investigates the effect of the ascomycetous fungus Trichoderma harzianum on lipid peroxidation, phospholipids
Jeong Kuk Park et al.
IUCrJ, 6(Pt 4), 729-739 (2019-07-19)
Ependymin was first discovered as a predominant protein in brain extracellular fluid in fish and was suggested to be involved in functions mostly related to learning and memory. Orthologous proteins to ependymin called ependymin-related proteins (EPDRs) have been found to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service