Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM025

Supelco

Trace Metals - Sandy Loam 8

certified reference material, pkg of 50 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

CofA

current certificate can be downloaded

packaging

pkg of 50 g

manufacturer/tradename

RTC CRM025-050

technique(s)

AAS: suitable
ICP: suitable

pH

7.21

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
This product was produced and certified from a naturally contaminated matrix and has not been fortified or modified in any way. Analytical data for Certification was obtained using USEPA SW846, 3rd edition methods 3050 (hot block) and 3051(microwave) using nitric acid extraction. Analysis was carried out according to USEPA methods 6010 (ICP-ES), 6020 (ICP-MS) and 7000 (AES).

Other Notes

The following elements are not certified and are included for information only:
  • Sb, B, Mo, Tl
Values of analytes vary lot to lot. Some lots may not contain all analytes.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Analyte

Description

    Aluminum
    Antimony
    Arsenic
    Barium
    Beryllium
    Boron
    Cadmium
    Calcium
    Chromium
    Cobalt
    Copper
    Iron
    Lead
    Magnesium
    Manganese
    Mercury
    Molybdenum
    Nickel
    Potassium
    Selenium
    Silicon
    Silver
    Sodium
    Thallium
    Vanadium
    Zinc
    See All (26)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1B - Skin Sens. 1

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service