Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M052080

Sigma-Aldrich

DMT-dI Phosphoramidite

configured for PerkinElmer, configured for Polygen

Synonym(s):

DMT-dI amidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H47N6O7P
CAS Number:
Molecular Weight:
754.81
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

product line

Proligo Reagents

Assay

≥98% (31P-NMR)
≥98.0% (reversed phase HPLC)

λ

conforms (UV/VIS Identity)

compatibility

configured for PerkinElmer
configured for Polygen

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)n3cnc4C(=O)NC=Nc34)(c5ccccc5)c6ccc(OC)cc6

InChI

1S/C40H47N6O7P/c1-27(2)46(28(3)4)54(51-22-10-21-41)53-34-23-36(45-26-44-37-38(45)42-25-43-39(37)47)52-35(34)24-50-40(29-11-8-7-9-12-29,30-13-17-32(48-5)18-14-30)31-15-19-33(49-6)20-16-31/h7-9,11-20,25-28,34-36H,10,22-24H2,1-6H3,(H,42,43,47)/t34-,35+,36+,54?/m0/s1

InChI key

ZKGZROFWRPJVEB-XZYUMQGLSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service