Skip to Content
MilliporeSigma
All Photos(1)

Documents

L1011

Sigma-Aldrich

Labetalol hydrochloride

>98% (TLC), powder

Synonym(s):

2-Hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]benzamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24N2O3 · HCl
CAS Number:
Molecular Weight:
364.87
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

>98% (TLC)

form

powder

color

white

solubility

H2O: soluble
ethanol: soluble

originator

Schering Plough

SMILES string

Cl.CC(CCc1ccccc1)NCC(O)c2ccc(O)c(c2)C(N)=O

InChI

1S/C19H24N2O3.ClH/c1-13(7-8-14-5-3-2-4-6-14)21-12-18(23)15-9-10-17(22)16(11-15)19(20)24;/h2-6,9-11,13,18,21-23H,7-8,12H2,1H3,(H2,20,24);1H

InChI key

WQVZLXWQESQGIF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Competitive β-adrenoceptor antagonist.

Features and Benefits

This compound was developed by Schering Plough. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S C Fagan et al.
Experimental neurology, 150(1), 153-158 (1998-03-26)
We examined the relationship between acute hypertension following cerebral embolization and subsequent hemorrhagic transformation (HT) in a rabbit embolic stroke model. We have shown previously that the likelihood and severity of hemorrhage were significantly correlated with the magnitude of an
R Jaboureck-Bouttier et al.
Hypertension in pregnancy, 18(3), 239-247 (1999-12-10)
Increased lipid peroxidation is a putative causal factor for preeclampsia. Because oxygen free radicals are involved in inducing the lipid oxidation chain reaction, we evaluated two beta adrenoreceptor blockers, labetalol and metoprolol, currently used for treating hypertension with regard to
E Riva et al.
British journal of pharmacology, 104(4), 823-828 (1991-12-01)
1. We compared the alpha 1-, alpha 2- and beta 1-adrenoceptor blocking potencies of labetalol with those of its two stereoisomers (RR and SR) in pithed rats and in homogenized rat cerebral cortex and heart. 2. In pithed rats, labetalol
Cheryl Brandenburg et al.
Frontiers in cellular neuroscience, 14, 577858-577858 (2020-11-27)
Autism spectrum disorder (ASD) is behaviorally defined and diagnosed by delayed and/or impeded language, stereotyped repetitive behaviors, and difficulties with social interactions. Additionally, there are disruptions in motor processing, which includes the intent to execute movements, interrupted/inhibited action chain sequences
Mohamed O Amin et al.
Mikrochimica acta, 186(8), 503-503 (2019-07-05)
The use of carbon black-Fe3O4 magnetic nanocomposite (CB-Fe3O4) as a probe for surface-enhanced laser desorption ionization mass spectrometry (SELDI-MS) with a high extraction efficiency and sensitive detection is described. The magnetic nanocomposite was synthesized and fully characterized using X-ray photoelectron

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service