Skip to Content
MilliporeSigma

P0884

(±)-Propranolol hydrochloride

≥99% (TLC), powder, β-adrenoceptor antagonist

Synonym(s):

(±)-1-Isopropylamino-3-(1-naphthyloxy)-2-propanol hydrochloride, DL-Propranolol hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H21NO2 · HCl
CAS Number:
Molecular Weight:
295.80
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
206-268-7
MDL number:
Beilstein/REAXYS Number:
4164259
Assay:
≥99% (TLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

(±)-Propranolol hydrochloride, ≥99% (TLC), powder

Quality Segment

assay

≥99% (TLC)

form

powder

color

white

solubility

ethanol: 10 mg/mL, DMSO: <14.5 mg/mL, H2O: 50 mg/mL, 45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 8.0 mg/mL

originator

AstraZeneca

storage temp.

2-8°C

SMILES string

Cl.CC(C)NCC(O)COc1cccc2ccccc12

InChI

1S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18H,10-11H2,1-2H3;1H

InChI key

ZMRUPTIKESYGQW-UHFFFAOYSA-N

General description

dissolve with heat. Aqueous solutions are most stable at pH 3.0 and decompose rapidly at basic pH. Decomposition is accompanied by discoloration of the solution.
Propranolol hydrochloride is easily soluble in ethanol and water.

Application

(±)-Propranolol hydrochloride has been used:
  • to determine whether autonomic nervous system mediated the promotive effect on skin microcirculation in rat
  • as a non-selective β- receptor blocker to reduce the arrhythmogenic events
  • to study its effect on oxygen-induced retinopathy in mice

Biochem/physiol Actions

Propranolol hydrochloride is a β-adrenoceptor antagonist. Its action at β2 receptor results in bronchoconstriction. Due to its lipophilic nature, propranolol can penetrate to the central nervous system and has a negative effect. It serves as a 5-HT1/5-HT2 serotonin receptor antagonist. Propranolol hydrochloride is useful as an antihypertensive drug, cardiac depressant and also in the treatment of angina pectoris. It decreases the effect of stress and exercise on heart by reducing the rate of contraction and conduction of impulse. It is known to competitively block the action of catecholamines.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the β-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Disclaimer

Shelf-life of the solid is at least four years if stored protected from the uptake of moisture.


Still not finding the right product?

Explore all of our products under (±)-Propranolol hydrochloride


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library