Skip to Content
MilliporeSigma
All Photos(2)

Documents

L0881

Sigma-Aldrich

Lectin from Arachis hypogaea (peanut)

lyophilized powder, Affinity-purified

Synonym(s):

PNA, Peanut agglutinin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

form

lyophilized powder

Quality Level

potency

<0.1 μg per mL agglutination activity

quality

Affinity-purified

impurities

salt, free

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Lectin from Arachis hypogaea (peanut) is a homotetrameric protein of 110 kDa.

Application

Lectin from Arachis hypogaea (peanut) has been used:
  • to detect Thomson-Friedenreich antigen
  • as a component in PBS to study its effects on cell viability
  • to study its ability to block the binding site

Biochem/physiol Actions

Lectin from Arachis hypogaea (peanut) possess specificity towards β -D-Gal(1-3)-D-galNAc residue of Thomson − Friedenreich antigen. It is the most widely used diagnostic and prognostic marker for many squamous cell carcinomas.
PNA does not agglutinate normal human erythrocytes, but strongly agglutinates neuraminidase treated erythrocytes. PNA has potent anti-T activity similar to the anti-T antibody in human sera. The lectin can be used to distinguish between human lymphocyte subsets.

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, of a 1 mg/ml solution. This activity is the lowest concentration to agglutinate a 2% suspension of neuraminidase treated human blood group O erythrocytes after 1 hr incubation at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adhesion of plant roots to poly-L-lysine coated polypropylene substrates
Towler MJ and Weathers PJ
Journal of Biotechnology, 101(2), 147-155 (2003)
Alejandro J Cagnoni et al.
Acta crystallographica. Section D, Structural biology, 76(Pt 11), 1080-1091 (2020-11-03)
Carbohydrate-lectin interactions are involved in important cellular recognition processes, including viral and bacterial infections, inflammation and tumor metastasis. Hence, structural studies of lectin-synthetic glycan complexes are essential for understanding lectin-recognition processes and for the further design of promising chemotherapeutics that
In vivo tumor cell adhesion in the pulmonary microvasculature is exclusively mediated by tumor cell-endothelial cell interaction
Gassmann P
BMC Cancer, 10(1), 177-177 (2010)
Lectin binding and effects in culture on human cancer and non-cancer cell lines: examination of issues of interest in drug design strategies
Petrossian K, et al.
Acta Histochemica, 109(6), 491-500 (2007)
Peter Gassmann et al.
BMC cancer, 10, 177-177 (2010-05-04)
Metastasis formation is the leading cause of death among colon cancer patients. We established a new in-situ model of in vivo microscopy of the lung to analyse initiating events of metastatic tumor cell adhesion within this typical metastatic target of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service