Skip to Content
MilliporeSigma
All Photos(1)

Documents

HYS010

Sigma-Aldrich

HyStem® Cell Culture Scaffold Kit

Trial Kit, For 2.5 mL of hydrogel scaffold solution

Synonym(s):

Cell Culture Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Customizable and free of animal components, Hystem hydrogel cell culture scaffolds provide a complex, three dimensional environment in which cells are able to proliferate, much as they would in vivo.
  • Customizable - each distinct cell type in a multicellular organism has its own unique microenvironment. Hystem offers the flexibility to tailor the environments of cultured cells to mimic their in vivo counterparts and optimize proliferation and differentiation.
  • Animal Component Free - As a synthetic Hyaluronic acid-based matrix, Hystem offers complete control of the cellular environment. All components are well-characterized, consistent, and derived from non-animal sources
  • Biologically accurate - Rich in hyaluronic acid, the Hystem hydrogel closely mimics the complex three dimensional cellular environments found in life, providing an optimal environment for culturing your cells.
The Hystem Trial Kit enables researchers to evaluate the Hystem technology in their own hands and confirm that Hystem is the right choice for their research.

With Hystem, researchers can tailor the incorporation of ECM proteins, cell attachment factors, and growth factors as well as the elasticity of the hydrogel and cell incorporation (encapsulations or top plating) to create the specific 3D environment required by their cells.

Hystem has been successfully used to expand human embryonic stem cells (H9s), umbilical cord blood CD23+ stem cells, hepatic stem cells and hepatic progenitor cells.

Hystem is recommended for in vivo experimentation.

Hystem offers a minimal number of cell attachment sites. If desired, researchers may incorporate attachment factors of their choice.

Legal Information

Hystem is a registered trademark of BIOTIME, INC

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Li et al.
Analytical chemistry, 92(3), 2794-2801 (2020-01-15)
Here, we develop an injection molded microfluidic approach for single cell analysis by making use of (1) rapidly curing injectable hydrogels, (2) a high density microfluidic weir trap array, and (3) reversibly bonded PDMS lids that are strong enough to

Articles

Hystem 3D hydrogels are based on hyaluronic acid, a major component of the extracellular matrix. 3D cell culture can be done using Hystem hydrogels.

Hystem 3D hydrogels are based on hyaluronic acid, a major component of the extracellular matrix. 3D cell culture can be done using Hystem hydrogels.

Hystem 3D hydrogels are based on hyaluronic acid, a major component of the extracellular matrix. 3D cell culture can be done using Hystem hydrogels.

Heparin is distinct from heparan sulfate proteoglycans (HS) which is primarily produced by mast cells. While, HS is produced by all cell types. Know more about the difference between heparin and heparin sulfate proteoglycan and select the right component for your study.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service