Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

M1073

Sigma-Aldrich

MaxGel ECM

96 well plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71
size:
96 wells

Quality Level

concentration

0.8-1.2 mg/mL protein

size

96 wells

suitability

suitable for (cell culture)

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

Application

This product creates a three-dimensional environment that improves cellular proflieration by promoting cell growth and migration, and has been shown to aid the proliferation of many types of cells, including human embryonic stem cells, neural stem cells, neurons, glia, astrocytes, fibroblasts, hepatocytes, and keratinocytes.

Components

MaxGel ECM contains human extracellular matrix components including collagens, laminin, fibronectin, tenascin, elastin and a number of proteoglycans and glycosaminoglycans. This basement membrane extract reproduces the cooperative interactions between epithelia and mesenchyme that occur during the development and organotypic cell culture of skin.

Caution

MaxGel will remain stable for at least 1 year when stored at -70°C.

Other Notes

Approximate volume per well is 75 μl

Legal Information

MaxGel is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne Tristan et al.
PloS one, 4(4), e5042-e5042 (2009-04-07)
Staphylococcus aureus necrotizing pneumonia is a severe disease caused by S. aureus strains carrying the Panton Valentine leukocidin (PVL) genes (lukS-PV & lukF-PV) encoded on various bacteriophages (such as phiSLT). Clinical PVL+ strains isolated from necrotizing pneumonia display an increased

Articles

Human cell derived basement membrane extract (BME) hydrogel for 3D cell culture of primary cell, stem cell and xenograph cancer models.

Hydrogela are the most widely used systems for 3D cell culture. Learn more about this technology (what are hydrogels? How to chose?)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service