Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HS1002B

Autoclave disposal bags

H × W × thickness 600 mm × 415 mm × 2.0 mm, transparent polypropylene (with blue biohazard labeling )

Synonym(s):

autoclave waste disposal, biohaz waste bag, biohazardous disposal bags, waste bags

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NB.17

material

transparent polypropylene (with blue biohazard labeling )

feature

label blue biohazard labeling

manufacturer/tradename

Heathrow Scientific HS1002B

H × W × thickness

600 mm × 415 mm × 2.0 mm

Looking for similar products? Visit Product Comparison Guide

General description

Autoclave Bags are clear, imprinted with blue biohazard labeling designed for safe disposal.
Constructed of 2.0 mm thick polypropylene, allowing for repeated autoclaving
  • Strong side seams for securely holding items
  • Front write-on panel
  • Autoclavable at 250°F (121°C).
  • 4 inner packs of 50 bags, Sold as a case of 200
  • Disposable

Autoclave disposal bags are clear, imprinted with blue biohazard labeling designed for safe disposal. It is made of 2.0 mm thick polypropylene and can be autoclaved multiple times.

Features and Benefits

  • Strong side seams for securing objects
  • Front write-on panel
  • Autoclavable at 250°F (121°C).
  • 4 inner packs of 50 bags in a case of 200

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service