Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.17
UNSPSC Code:
41106514
material
transparent polypropylene (with blue biohazard labeling )
feature
label blue biohazard labeling
manufacturer/tradename
Heathrow Scientific HS1002C
H × W × thickness
810 mm × 610 mm × 2.0 mm
General description
Autoclave Bags are clear, imprinted with blue biohazard labeling designed for safe disposal.
Constructed of 2.0 mm thick polypropylene, allowing for repeated autoclaving
Constructed of 2.0 mm thick polypropylene, allowing for repeated autoclaving
- Strong side seams for securely holding items
- Front write-on panel
- Autoclavable at 250°F (121°C).
- 4 inner packs of 50 bags, Sold as a case of 200
- Disposable
Autoclave disposal bags are clear, imprinted with blue biohazard labeling designed for safe disposal. It is made of 2.0 mm thick polypropylene and can be autoclaved multiple times.
Features and Benefits
- Strong side seams for securing objects
- Front write-on panel
- Autoclavable at 250°F (121°C).
- 4 inner packs of 50 bags in a case of 200
Still not finding the right product?
Explore all of our products under Autoclave disposal bags
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| HS1002C | 04061835302123 |