MilliporeSigma
HS10320
Biohazard disposal bags
H × W × thickness 305 mm × 203 mm × 0.04 mm, red polypropylene (with biohazard warning symbol & precautions in English, Spanish, & French)
Synonym(s):
autoclave waste disposal, biohaz waste bag, biohazardous disposal bags, waste bags
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.17
UNSPSC Code:
41106514
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
red polypropylene (with biohazard warning symbol & precautions in English, Spanish, & French)
feature
label
packaging
pkg of 500 ea
manufacturer/tradename
Heathrow Scientific HS10320
H × W × thickness
305 mm × 203 mm × 0.04 mm
General description
Biohazard disposal bags are printed with biohazard warning symbols. A wide range of biohazard collection bags with sizes to suit all needs are available for personal or general lab use.
For personal or general lab use, sizes to suit all needs
A range of biohazard collection bags printed with biohazard warning symbol and precautions in English, Spanish, and French
  • Includes an indicator patch that darkens when autoclaved
  • Autoclavable to 250°F (121°C)
  • Disposable
  • Epoxy coated steel wire holder available HS10044H
Features and Benefits
  • Two thicknesses of polypropylene are available
  • An indicator patch that darkens when autoclaved is included
  • Autoclavable to 250°F (121°C)
  • Disposable
  • An epoxy-coated steel wire holder is available (HS10044H).
Still not finding the right product?
Explore all of our products under Biohazard disposal bags
Choose from one of the most recent versions:
Certificates of Analysis (COA)