Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA015037

Sigma-Aldrich

Anti-ARID5B antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ARID domain- containing protein 5B, Anti-AT-rich interactive domain-containing protein 5B, Anti-MRF-2, Anti-Modulator recognition factor 2, Anti-Mrf1-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

TSKYPSRDMYRESENSSFPSHRHQEKLHVNYLTSLHLQDKKSAAAEAPTDDQPTDLSLPKNPHKPTGKVLGLAHSTTGPQESKGISQFQVLGSQSRDCHPKACRVSPMTMSGPKKYPESLSRSGKPHHVRLENFRKME

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARID5B(84159)

General description

ARID5B (AT rich interactive domain 5B) belongs to the family of DNA-binding proteins called AT-rich interaction domain (ARID). This gene is localized to human chromosome 10q21.2. This protein contains three ARID domains, which includes six α-helices in the central region, called H1 to H6. The other two domains are called L1 and L2, which are DNA-binding loops.

Immunogen

AT-rich interactive domain-containing protein 5B recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-ARID5B antibody produced in rabbit has been used in chromatin immunoprecipitation (ChIP) and western blotting.

Biochem/physiol Actions

ARID5B (AT rich interactive domain 5B) controls adipogenesis and liver development, by regulating the transcription of involved genes. It interacts with DNA by binding with PHD finger protein 2, to form a histone H3K9me2 demethylase complex. This gene influences the development and differentiation of hematopoietic cells, and variants in this gene are linked with childhood acute lymphoblastic leukemia (ALL). Studies in Spanish population show that intron 3 of this gene is a hot spot for susceptibility to acute lymphoblastic leukemia. It also strongly influences cell growth and differentiation, and the development of embryo.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72808

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chien-Yu Lin et al.
Blood cells, molecules & diseases, 52(2-3), 140-145 (2013-11-10)
Childhood acute lymphoblastic leukemia (ALL), a heterogeneous disease that includes multiple subtypes is defined by cell lineage and chromosome anomalies. Previous genome-wide association studies have reported several ARID5B and IKZF1 single nucleotide polymorphisms (SNPs) associated with the incidence of ALL.
Important Role of FTO in the Survival of Rare Panresistant Triple-Negative Inflammatory Breast Cancer Cells Facing a Severe Metabolic Challenge
Singh B, et al.
PLoS ONE, 11(7), e0159072-e0159072 (2016)
ARID5B as a critical downstream target of the TAL1 complex that activates the oncogenic transcriptional program and promotes T-cell leukemogenesis
Leong WZ, et al.
Genes & Development, 31(23-24), 2343-2360 (2017)
Ángela Gutiérrez-Camino et al.
Journal of cancer research and clinical oncology, 139(11), 1879-1886 (2013-09-10)
Single-nucleotide polymorphisms (SNPs) in AT-rich interactive domain 5B (ARID5B) have been associated with risk for pediatric acute lymphoblastic leukemia (ALL). After reviewing previous studies, we realized that the most significant associations were restricted to intron 3, but the mechanism(s) by
Sheng Cai et al.
Biochemistry, 46(17), 4943-4950 (2007-04-05)
Understanding the mechanism of protein-DNA interactions at the molecular level is one of the main focuses in structural and molecular biological investigations. At present, NMR spectroscopy is the only approach that can provide atomic details of protein-DNA recognition in solution.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service